Skip to Content
Merck
All Photos(1)

Documents

530603

Sigma-Aldrich

2,5-Diaminopyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3
CAS Number:
Molecular Weight:
109.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

bp

180 °C/12 mmHg (lit.)

SMILES string

Nc1ccc(N)nc1

InChI

1S/C5H7N3/c6-4-1-2-5(7)8-3-4/h1-3H,6H2,(H2,7,8)

InChI key

MIROPXUFDXCYLG-UHFFFAOYSA-N

General description

2,5-Diaminopyridine is a disubstituted pyridine that can be prepared by the reduction of 5-nitro-2-aminopyridine using 10%Pd/C.

Application

2,5-Diaminopyridine may be used in the preparation of 6-acetamidoimidazo[ 1,2-a]pyridine. It may be used in the preparation of organo-soluble polyimides by reacting with different dianhydride monomers. These polyimides form the precursor for preparing novel polyimide-silica composites.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porous pyridine based polyimide?silica nanocomposites with low dielectric constant.
K?v?lc?m N, et al.
Journal of Porous Materials, 20(4), 709-718 (2013)
Teleamination of the imidazo [1,2-a] pyridine system.
Hand ES and Paudler WW.
The Journal of Organic Chemistry, 43(14), 2900-2906 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service