Skip to Content
Merck
All Photos(1)

Key Documents

529567

Sigma-Aldrich

Cadmium perchlorate hydrate

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(ClO4)2·xH2O
CAS Number:
Molecular Weight:
311.31 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

crystals and lumps

composition

Degree of hydration, ~6

reaction suitability

reagent type: catalyst
core: cadmium

reagent type: oxidant

impurities

≤15.0 ppm Trace Metal Analysis

SMILES string

O.[Cd++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/Cd.2ClHO4.H2O/c;2*2-1(3,4)5;/h;2*(H,2,3,4,5);1H2/q+2;;;/p-2

InChI key

SGAGEPDIZCCCPW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Ox. Sol. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baoyan Xing et al.
ChemistryOpen, 9(10), 1018-1026 (2020-10-20)
Solid-state NMR analysis on wurtzite alloyed CdSe1-xSx crystalline nanoparticles and nanobelts provides evidence that the 113Cd NMR chemical shift is not affected by the varying sizes of nanoparticles, but is sensitive to the S/Se anion molar ratios. A linear correlation
Siyi Hu et al.
The Analyst, 139(18), 4681-4690 (2014-07-24)
Biofunctionalized quantum dots (QDs), especially protein-coated QDs, are known to be useful targeted fluorescent labels for cellular and deep-tissue imaging. These nanoparticles can also serve as efficient energy donors in fluorescence resonance energy transfer (FRET) binding assays for the multiplexed
Herman S Mansur et al.
Materials science & engineering. C, Materials for biological applications, 52, 61-71 (2015-05-09)
Cancer remains one of the world's most devastating diseases with millions of fatalities and new cases every year. In this work, we attempted to develop a facile "enzyme-free" fluoroimmunoassay based on the novel nanoconjugates composed of CdS quantum dots (QDs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service