Skip to Content
Merck
All Photos(1)

Documents

526169

Sigma-Aldrich

1,8-Dimethyl-1,4,8,11-tetraazacyclotetradecane

Synonym(s):

1,8-Dimethylcyclam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H28N4
CAS Number:
Molecular Weight:
228.38
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.4940 (lit.)

Quality Level

bp

98-99 °C (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

CN1CCCNCCN(C)CCCNCC1

InChI

1S/C12H28N4/c1-15-9-3-5-14-8-12-16(2)10-4-6-13-7-11-15/h13-14H,3-12H2,1-2H3

InChI key

BNLDMZVBFXARKJ-UHFFFAOYSA-N

General description

1,8-Dimethyl-1,4,8,11-tetraazacyclotetradecane is a heterocyclic building block.

Application

1,8-Dimethyl-1,4,8,11-tetraazacyclotetradecane may be used as a precursor for the synthesis of `trans′-difunctionalized derivatives. It may also be used in the synthesis of 4,11-bis(N-pyren-1-yl-acetamide)-1,8-dimethyl-1,4,8,11-tetraazacyclotetradecane.
Reactant for:
  • Ferromagnetic coupling of copper(II) and nickel(II) complexes
  • Preparation of cyclam bridged dinuclear platinum(II) complex as an antitumor agent
  • Preparation of dimethylcyclam based fluoroionophore having Hg2+- and Cd2+-selective signaling behaviors

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.1 °F - closed cup

Flash Point(C)

90.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

So-Youn Moon et al.
The Journal of organic chemistry, 70(6), 2394-2397 (2005-03-12)
[reaction: see text] A new fluoroionophore has been synthesized by appending two signaling pyrenylacetamide subunits on the binding motif of 1,8-dimethylcyclam. The designed compound exhibited highly selective and sensitive fluoroionophoric behavior toward Hg(2+) ions of excimer emission in aqueous dioxane
Copper (II) and Nickel (II) Complexes oftrans'-Difunctionalized Tetraaza Macrocycles.
Comparone A and Kaden TA.
Helvetica Chimica Acta, 81(10), 1765-1772 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service