Skip to Content
Merck
All Photos(2)

Key Documents

52605

Sigma-Aldrich

Hexamethonium hydroxide solution

~0.1 M in H2O

Synonym(s):

N,N,N,N′,N′,N′-Hexamethylhexamethylenediammonium dihydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(OH)(CH2)6N(OH)(CH3)3
CAS Number:
Molecular Weight:
236.39
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

concentration

~0.1 M in H2O

impurities

≤0.01% halides (as bromide)

functional group

amine

storage temp.

2-8°C

SMILES string

[OH-].[OH-].C[N+](C)(C)CCCCCC[N+](C)(C)C

InChI

1S/C12H30N2.2H2O/c1-13(2,3)11-9-7-8-10-12-14(4,5)6;;/h7-12H2,1-6H3;2*1H2/q+2;;/p-2

InChI key

GYLUMIIRFKDCKI-UHFFFAOYSA-L

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Akerman et al.
Brain : a journal of neurology, 135(Pt 12), 3664-3675 (2012-10-16)
Trigeminal autonomic cephalalgias are highly disabling primary headache disorders, characterized by severe unilateral head pain and associated ipsilateral cranial autonomic features. There is limited understanding of their pathophysiology and how and where treatments act to reduce symptoms; this is significantly
Hiroshi Hanamoto et al.
International journal of oral science, 4(1), 24-29 (2012-03-06)
Stimulation of the trigeminal nerve can elicit various cardiovascular and autonomic responses; however, the effects of anesthesia with pentobarbital sodium on these responses are unclear. Pentobarbital sodium was infused intravenously at a nominal rate and the lingual nerve was electrically
Kamila Shakarchi et al.
The Journal of experimental biology, 216(Pt 5), 869-880 (2012-11-17)
The chemosensory roles of gill neuroepithelial cells (NECs) in mediating the hyperventilatory response to hypoxia are not clearly defined in fish. While serotonin (5-HT) is the predominant neurotransmitter in O(2)-sensitive gill NECs, acetylcholine (ACh) plays a more prominent role in
Wen-Hsien Lu et al.
Critical care medicine, 41(5), 1276-1285 (2013-02-08)
Among enterovirus 71 infections, brainstem encephalitis progressing abruptly to cardiac dysfunction and pulmonary edema causes rapid death within several hours. However, no currently known early indicators and treatments can monitor or prevent the unexpectedly fulminant course. We investigate the possible
Wet lungs and a battered brain stem: can we stop one if we cannot stop the other?
Paul Nyquist
Critical care medicine, 41(5), 1373-1374 (2013-04-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service