Skip to Content
Merck
All Photos(2)

Key Documents

523690

Sigma-Aldrich

3-Amino-1-adamantanol

96%

Synonym(s):

(3-Hydroxyadamantan-1-yl)amine, 1-Amino-3-adamantanol, 1-Amino-3-hydroxyadamantane, 3-Amino-1-hydroxyadamantane, 3-Aminotricyclo[3.3.1.13,7]decan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO
CAS Number:
Molecular Weight:
167.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

265 °C (dec.) (lit.)

functional group

hydroxyl

SMILES string

N[C@]12C[C@@H]3C[C@H](C1)C[C@@](O)(C3)C2

InChI

1S/C10H17NO/c11-9-2-7-1-8(3-9)5-10(12,4-7)6-9/h7-8,12H,1-6,11H2/t7-,8+,9+,10-

InChI key

DWPIPTNBOVJYAD-FIRGSJFUSA-N

Application

Employed in the synthesis of inhibitors with antihyperglycemic properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NOVEL TRICYCLIC CYANOPYRROLIDINE DERIVATIVES AS DPP4 INHIBITORS: AN IMPROVED SYNTHESIS OF TRICYCLIC a-CYCNOPYRROLIDINE FROM CAMPHOR.
Arumugam K and Anitha M.
Rasayan Journal of Chemistry, 6(3), 230-237 (2013)
A Facile and Economical Method to Synthesize Vildagliptin.
Deng Y, et al.
Letters in Organic Chemistry, 11(10), 780-784 (2014)
Nieves Fresno et al.
PloS one, 9(12), e113841-e113841 (2014-12-02)
Paracetamol also known as acetaminophen, is a widely used analgesic and antipyretic agent. We report the synthesis and biological evaluation of adamantyl analogues of paracetamol with important analgesic properties. The mechanism of nociception of compound 6a/b, an analog of paracetamol
Edwin B Villhauer et al.
Journal of medicinal chemistry, 46(13), 2774-2789 (2003-06-13)
Dipeptidyl peptidase IV (DPP-IV) inhibition has the potential to become a valuable therapy for type 2 diabetes. The synthesis and structure-activity relationship of a new DPP-IV inhibitor class, N-substituted-glycyl-2-cyanopyrrolidines, are described as well as the path that led from clinical
Ramzia I El-Bagary et al.
International journal of biomedical science : IJBS, 7(3), 201-208 (2011-09-01)
Two reversed-phase liquid chromatographic (RP-LC) methods are described for the determination of two binary mixtures of hypoglycemic agents. In the first method, vildagliptin (VDG) was determined in the presence of 3-amino-1-adamantanol (AAD), a synthetic intermediate and impurity of VDG. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service