Skip to Content
Merck
All Photos(1)

Documents

513059

Sigma-Aldrich

Ethyl 3,4,5-trimethoxybenzoylacetate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2COCH2CO2C2H5
CAS Number:
Molecular Weight:
282.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

88-91 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1cc(OC)c(OC)c(OC)c1

InChI

1S/C14H18O6/c1-5-20-13(16)8-10(15)9-6-11(17-2)14(19-4)12(7-9)18-3/h6-7H,5,8H2,1-4H3

InChI key

VEYNISLTHXQSGU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethyl 3, 4, 5-Trimethoxybenzoylacetate.
Anderson Jr AG and Greef HF.
Journal of the American Chemical Society, 74(11), 2923-2923 (1952)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 327-327 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service