Skip to Content
Merck
All Photos(1)

Key Documents

495344

Sigma-Aldrich

Poly(3-decylthiophene-2,5-diyl)

regioregular, average Mw ~42,000, average Mn ~30,000

Synonym(s):

P3DT, P3DT-RR

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C14H22S)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

mol wt

average Mn ~30,000
average Mw ~42,000

color

black

fluorescence

λex 441 nm; λem 567 nm in chloroform

SMILES string

[s]1cc(cc1)CCCCCCCCCC

InChI key

JAYBIBLZTQMCAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3-decylthiophene-2,5-diyl) (P3DT) is a conducting polymer with excellent electrical and optical properties. It is soluble in most of the organic solvents and can be used in a variety of electrochemical applications.

Application

Conducting polymer. Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.
P3DT is a conjugated polymer that can be used for a variety of applications, which include anticorrosive additives in paints, organic photovoltaic cells, polymeric solar cells, and field effect transistors.
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.

Packaging

Packaged in glass bottles

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The thermal behaviour of low-molecular-weight poly (3-decylthiophene)
Bolognesi A, et al.
Die Makromolekulare Chemie, 194(3), 817-827 (1993)
Poly (3-alkylthiophene) s as anticorrosive additive for paints: Influence of the main chain stereoregularity
Iribarren J, et al.
Journal of Applied Polymer Science, 108(5), 3291-3297 (2008)
Field-effect mobility of charge carriers in blends of regioregular poly (3-alkylthiophene) s
Babel A and Jenekhe SA
The Journal of Physical Chemistry B, 107(8), 1749-1754 (2003)
Application of a polythiophene derivative as anticorrosive additive for paints
Ocampo C, et al.
Progress in Organic Coatings, 53(3), 217-224 (2005)
Poly (3-decylthiophene) radical anions and cations in solution: Single and multiple polarons and their delocalization lengths in conjugated polymers
Takeda N and Miller JR
The Journal of Physical Chemistry B, 116(50), 14715-14723 (2012)

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service