Skip to Content
Merck
All Photos(1)

Key Documents

494011

Sigma-Aldrich

(4-Bromophenylethynyl)trimethylsilane

98%

Synonym(s):

(p-Bromophenyl)ethynyl)trimethylsilane, 1-(p-Bromophenyl)-2-trimethylsilylacetylene, 1-Bromo-4-(2-trimethylsilylethynyl)benzene, 1-Bromo-4-(trimethylsilylethynyl)benzene, 1-Bromo-4-[2-(trimethylsilyl)ethynyl]benzene, 2-(p-Bromophenyl)-1-trimethylsilylacetylene, 4-(Trimethylsilylethynyl)phenyl bromide, [(4-Bromophenyl)ethynyl]trimethylsilane, [1-(4-Bromophenyl)-2-trimethylsilyl]acetylene, [2-(4-Bromophenyl)ethynyl]trimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4C≡CSi(CH3)3
CAS Number:
Molecular Weight:
253.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

61-63 °C (lit.)

functional group

bromo

SMILES string

C[Si](C)(C)C#Cc1ccc(Br)cc1

InChI

1S/C11H13BrSi/c1-13(2,3)9-8-10-4-6-11(12)7-5-10/h4-7H,1-3H3

InChI key

RNMSGCJGNJYDNS-UHFFFAOYSA-N

General description

(4-Bromophenylethynyl)trimethylsilane can be synthesized by the palladium catalyzed reaction between 4-bromo-1-iodobenzene and trimethylsilylacetylene. It undergoes Buchwald-Hartwig coupling with para-substituted diphenylamines.

Application

(4-Bromophenylethynyl)trimethylsilane may be used to synthesize:
  • 1-bromo-4-ethynylbenzene
  • 4-(4-bromophenyl)-3-butyn-2-one
  • 4-ethynyl-4′-tert-butylbiphenyl
  • 1,4-bis[2-(4-bromophenyl)ethynyl]-2,5-dihexylbenzene

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental and computational probes of the space in a self-assembled capsule.
Ajami D, et al.
Proceedings of the National Academy of Sciences of the USA, 103(24), 8934-8936 (2006)
Tunable, strongly-donating perylene photosensitizers for dye-sensitized solar cells.
Mathew S and Imahori H.
Journal of Materials Chemistry, 21(20), 7166-7174 (2011)
Synthesis and Solid-State Investigations of Oligo-Phenylene-Ethynylene Structures with Halide End-Groups.
Jenny NM, et al.
European Journal of Organic Chemistry, 2012(14), 2738-2747 (2012)
Probing surface-adlayer conjugation on organic-modified Si (111) surfaces with microscopy, scattering, spectroscopy, and density functional theory.
Kellar JA, et al.
The Journal of Physical Chemistry C, 113(7), 2919-2927 (2009)
Enantioselective synthesis of both enantiomers of various propargylic alcohols by use of two oxidoreductases.
Schubert T, et al.
European Journal of Organic Chemistry, 2001(22), 4181-4187 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service