493090
N-Butylpyridinium chloride-d14
98 atom % D
Synonym(s):
1-Butylpyridinium chloride-d14
About This Item
Recommended Products
isotopic purity
98 atom % D
form
solid
mp
131-133 °C (lit.)
mass shift
M+14
SMILES string
[Cl-].[2H]c1c([2H])c([2H])[n+](c([2H])c1[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
InChI
1S/C9H14N.ClH/c1-2-3-7-10-8-5-4-6-9-10;/h4-6,8-9H,2-3,7H2,1H3;1H/q+1;/p-1/i1D3,2D2,3D2,4D,5D,6D,7D2,8D,9D;
InChI key
POKOASTYJWUQJG-CBZSTONXSA-M
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service