Skip to Content
Merck
All Photos(1)

Key Documents

489751

Sigma-Aldrich

Imidazole-15N2

98 atom % 15N, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H415N2
CAS Number:
Molecular Weight:
70.06
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Assay

98% (CP)

form

solid

bp

256 °C (lit.)

mp

89-91 °C (lit.)

mass shift

M+2

SMILES string

c1c[15nH]c[15n]1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)/i4+1,5+1

InChI key

RAXXELZNTBOGNW-MQIHXRCWSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan-Bernd Hövener et al.
Angewandte Chemie (International ed. in English), 57(35), 11140-11162 (2018-02-28)
Magnetic resonance (MR) is one of the most versatile and useful physical effects used for human imaging, chemical analysis, and the elucidation of molecular structures. However, its full potential is rarely used, because only a small fraction of the nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service