Skip to Content
Merck
All Photos(3)

Documents

483745

Sigma-Aldrich

Sodium

cubes, contains mineral oil, 99.9% trace metals basis

Synonym(s):

Sodium metal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na
CAS Number:
Molecular Weight:
22.99
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 440 °C)

Quality Level

Assay

99.9% trace metals basis

form

cubes

contains

mineral oil

reaction suitability

reagent type: reductant

resistivity

4.69 μΩ-cm, 20°C

bp

883 °C (lit.)

mp

97.8 °C (lit.)

SMILES string

[Na]

InChI

1S/Na

InChI key

KEAYESYHFKHZAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium serves as a reductant for the difunctionalization of alkenes and alkynes. It is also used in Bouveault-Blanc reduction.

Application

Sodium metal can be used as a reducing agent in reactions such as Birch reduction.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Use of Sodium Metal in the Birch Reduction of Aromatic Compounds1.
Dryden Jr HL, et al.
The Journal of Organic Chemistry, 26(9), 3237-3245 (1961)
Recent advances in the use of sodium dispersion for organic synthesis
De PB
Synthesis, 53, 3180-3192 (2021)
Nicole Li et al.
American heart journal, 166(5), 815-822 (2013-11-02)
Cardiovascular diseases are the leading cause of death and disability in China. High blood pressure caused by excess intake of dietary sodium is widespread and an effective sodium reduction program has potential to improve cardiovascular health. This study is a
Supriya S Jalimarada et al.
Investigative ophthalmology & visual science, 54(6), 4330-4340 (2013-06-08)
Mutations in SLC4A11, a member of the SLC4 superfamily of bicarbonate transporters, give rise to corneal endothelial cell dystrophies. SLC4A11 is a putative Na⁺ borate and Na⁺:OH⁻ transporter. Therefore we ask whether SLC4A11 in corneal endothelium transports borate (B[OH]₄⁻), bicarbonate
Julijana Vuchkova et al.
American journal of physiology. Renal physiology, 307(5), F525-F532 (2014-07-06)
Albuminuria in nephrotic states is thought to arise from the formation of large pores in the glomerular capillary wall as large hydrodynamic probes, like Ficoll, have increased fractional clearance. In the present study, we tested for large pore formation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service