Skip to Content
Merck
All Photos(2)

Key Documents

482994

Sigma-Aldrich

[5,6]-Fullerene-C70

greener alternative

98%

Synonym(s):

Buckminsterfullerene C70, Fullerene-C70

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70
CAS Number:
Molecular Weight:
840.75
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>280 °C (lit.)

solubility

benzene: soluble
benzene: soluble
toluene: soluble
toluene: soluble

Orbital energy

HOMO 6.2 eV 
LUMO 4.6 eV 

greener alternative category

SMILES string

C1CC2C3CCC4C5C3C6C2C7C1CC8CC9CC%10CC%11C%12CC%13CC4C%13%14C%12C%15C%11C%16C%10C9C%17C8C7C%18C%17C%16C%15C(C6%18)C5%14

InChI

1S/C70/c1-2-22-5-6-24-13-14-26-11-9-23-4-3(21(1)51-52(22)54(24)55(26)53(23)51)33-31(1)61-35-7-8-27-15-16-29-19-20-30-18-17-28-12-10(25(7)56-57(27)59(29)60(30)58(28)56)37(35)63(33)65-36(4)40(9)67(44(17)42(12)65)69-46(11)47(14)70(50(20)49(18)69)68-43(13)39(6)66(45(16)48(19)68)64-34(5)32(2)62(61)38(8)41(15)64

InChI key

ATLMFJTZZPOKLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(5,6)-Fullerene-C70 (C70) is an ellipsoidal fullerene which has stable spheroidal carbon molecules.
C70 is an n-channel organic semiconductor. High reactivity of fullerenes is attributed to its characteristic pyramidalization of the sp2 carbons. Vacuum dried to remove residual solvent.Typical hydrocarbon content <0.25%.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

C70 may be incorporated into titanium oxide (TiO2) based nanowires and it forms a nanocomposite for light responsive photocatalyst. High performance organic photovoltaic (OPV) cells may be fabricated by using C70 as a photo-absorptive material.
C70 was used in the quantitative analysis of fullerenes in river sediment and soils.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annealing effect in the sexithiophene:C70 small molecule bulk heterojunction organic photovoltaic cells
Sakai, J.; Taima, T.; Yamanari, T.; et al.
Solar Energy Materials and Solar Cells, 93, 1149-1154 (2010)
Quantitative trace analysis of fullerenes in river sediment from Spain and soils from Saudi Arabia
Sanchis J, et al.
Analytical and Bioanalytical Chemistry, 405(18), 5915-5923 null
Benzyne Adds Across a Closed 5-6 Ring Fusion in C70: Evidence for Bond Delocalization in Fullerenes.
Meier MS, et al.
Journal of the American Chemical Society, 120, 2337-2342 (1998)
Annealing effect in the sexithiophene: C70 small molecule bulk heterojunction organic photovoltaic cells.
Sakai J, et al.
Solar Energy Materials and Solar Cells, 93(6-7), 1149-1153 (2009)
Sarah K Norton et al.
The Journal of allergy and clinical immunology, 130(3), 761-769 (2012-06-06)
Fullerenes are molecules being investigated for a wide range of therapeutic applications. We have shown previously that certain fullerene derivatives (FDs) inhibit mast cell (MC) function in vitro, and here we examine their in vivo therapeutic effect on asthma, a disease in

Articles

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Find various photovoltaic and bioscience-based applications of fullerenes.

Single-walled carbon nanotubes (SWCNTs) are promising materials for use in the active channel of field-effect transistors (FETs), photoabsorbing layers of solar cells and photodetectors because of their ultrafast charge transport mobility.

SWCNTs show promise in FETs, solar cells, and photodetectors due to their ultrafast charge transport mobility.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service