Skip to Content
Merck
All Photos(2)

Key Documents

476013

Sigma-Aldrich

Isopropyl isothiocyanate

97%

Synonym(s):

Isopropyl mustard oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNCS
CAS Number:
Molecular Weight:
101.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.493 (lit.)

bp

29-30 °C/10 mmHg (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

CC(C)N=C=S

InChI

1S/C4H7NS/c1-4(2)5-3-6/h4H,1-2H3

InChI key

VHBFEIBMZHEWSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropyl isothiocyanate is an isothiocyanate derivative. It has been identified as one of the volatile components in the following:
  • seed kernel and leaf of Moringa peregrina (Forssk.) Fiori, Agricolt1
  • essential oil isolated from Upland Wasabi
  • hydrodistillates of hedge mustard (Sysimbrium officinale)

Molecular conformations of isopropyl isothiocyanate in gas phase have been determined by electron diffraction studies. It has been synthesized by employing isopropyl amine as a starting reagent.

Application

Isopropyl isothiocyanate may be used to synthesize 4-bromo-5,6-dichloro-2-isopropylaminobenzimidazole.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of volatile components in essential oil of upland wasabi and their inhibitory effects on platelet aggregation.
Kumagai H, et al.
Bioscience, Biotechnology, and Biochemistry, 58(12), 2131-2135 (1994)
A general and facile one-pot process of isothiocyanates from amines under aqueous conditions.
Sun N, et al.
Beilstein Journal of Organic Chemistry, 8(1), 61-70 (2012)
Synthesis and antiviral evaluation of halogenated ?-d-and-l-erythrofuranosylbenzimidazoles.
Gudmundsson KS, et al.
Journal of Medicinal Chemistry, 43(12), 2464-2472 (2000)
Electron diffraction investigation of the molecular structures of isopropyl isocyanate and isopropyl isothiocyanate.
Cradock S, et al.
Journal of Molecular Structure, 140(3), 199-207 (1986)
Ivica Blazević et al.
Chemistry & biodiversity, 7(8), 2023-2034 (2010-08-24)
Volatile compounds of hedge mustard (Sysimbrium officinale) have been investigated for the first time. Forthy-two compounds were identified after hydrodistillation (without or upon autolysis) after gas chromatography and gas chromatography/mass spectrometry analyses. In addition, after decoction and hydrolysis of O-glycosides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service