Skip to Content
Merck
All Photos(1)

Key Documents

471984

Sigma-Aldrich

9-Oxabicyclo[6.1.0]non-4-ene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O
CAS Number:
Molecular Weight:
124.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.495 (lit.)

bp

195 °C (lit.)

density

1.013 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

C1CC2OC2CCC=C1

InChI

1S/C8H12O/c1-2-4-6-8-7(9-8)5-3-1/h1-2,7-8H,3-6H2/b2-1-

InChI key

YWFPXWMSGJXUFS-UPHRSURJSA-N

General description

9-Oxabicyclo[6.1.0]non-4-ene is an epoxide. It undergoes halofluorination reactions with N-halosuccinimides and triethylamine tris-hydrofluoride or Olah′s reagent.

Application

9-Oxabicyclo[6.1.0]non-4-ene may be used in the following studies:
  • novel bicyclo[4.2.1] and bicyclo[3.3.1] ethers
  • trans, trans-2,6-dibromo-9-oxabicyclo[3.3.1]nonane and trans, trans-2,5-dibromo-9-oxabicyclo[4.2.1]nonane
  • (Z)-2-(cyclooct-4-enyloxy)acetic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karl J Bonney et al.
The Journal of organic chemistry, 76(1), 97-104 (2010-12-01)
9-Oxabicyclo[6.1.0]non-4-ene (1) undergoes intramolecular bromonium ion-assisted epoxide ring-opening using N-bromosuccinimide via a presumed oxonium ion that is subject to stereospecific, nonregioselective capture with added external nucleophiles producing novel bicyclo[4.2.1] and bicyclo[3.3.1] ethers. Carboxylic acids (as catalyzed by tetramethylguanidine), alcohols, water
Stereoselective synthesis of cyclic ethers via bromine assistedepoxide ring expansion.
Davies SG, et al.
Tetrahedron Letters, 26(11), 1461-1464 (1985)
High-yielding, two-step 18F labeling strategy for 18F-PARP1 inhibitors.
Edmund J Keliher et al.
ChemMedChem, 6(3), 424-427 (2011-03-02)
Transannular oxygen participation in halofluorination reactions of 9-oxabicyclo [6.1. 0] non-4-ene [1].
Haufe G, et al.
Journal of Fluorine Chemistry, 46(1), 83-95 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service