458694
(Vinylbenzyl)trimethylammonium chloride
99%
Synonym(s):
(ar-Vinylbenzyl)trimethylammonium chloride
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
H2C=CHC6H4CH2N(CH3)3Cl
CAS Number:
Molecular Weight:
211.73
Beilstein:
3730835
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99%
mp
240 °C (dec.) (lit.)
SMILES string
[Cl-].C[N+](C)(C)Cc1ccc(C=C)cc1
InChI
1S/C12H18N.ClH/c1-5-11-6-8-12(9-7-11)10-13(2,3)4;/h5-9H,1,10H2,2-4H3;1H/q+1;/p-1
InChI key
TVXNKQRAZONMHJ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
(Vinylbenzyl)trimethylammonium chloride is a quaternary ammonium salt commonly used as a monomer in polymerization reactions to produce unique polymers. Its vinylbenzyl group allows for easy radical polymerization and copolymerization, enabling incorporation into different polymer backbones like polyacrylates, polyvinyl acetates, and styrene-based polymers. These resulting polymers possess excellent cationic properties, making them suitable for applications such as water treatment, papermaking, textiles, coatings, flocculants, coagulants, dispersants, binders, adhesives, and modifiers.
Application
(Vinylbenzyl)trimethylammonium chloride (VBTA) can be used as a monomer:
- To synthesize the poly(AMA-DVB-VBTA) monolith columns, which are used for solid-phase microextraction in analytical chemistry.
- To prepare an anion exchange membrane for a lignin oxidizing electrolyzer. VBTA provides the anion exchange functionality to the copolymer membrane, which allows for the selective transport of anions across the membrane
- Poly[oligo(ethylene glycol) methacrylate]-b-based nanostructures for encapsulating magnetic nanoparticles and DNA.
- Nano-sized polyelectrolyte complexes.
- Homo and block copolymers of poly(VBTMAC) for non-viral DNA delivery systems.
Other Notes
Contains a mixture of 3-vinyl isomers and 4-vinyl isomers
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Poly[oligo(ethylene glycol) methacrylate]-b-poly[(vinyl benzyl trimethylammonium chloride)] Based Multifunctional Hybrid Nanostructures Encapsulating Magnetic Nanoparticles and DNA
Angeliki Chroni, et al.
Polymers, 12 (2020)
Chen Zheng et al.
Polymers, 12(3) (2020-04-03)
By using different conductivity of polyaniline as filler, a kind of poly(ionic liquid)/polyaniline composite particles was synthesized to investigate the influence of dielectric polarization rate difference between filler and matrix on the electrorheological response and flow stability of composite-based electrorheological
Poly(vinyl benzyl trimethylammonium chloride) Homo and Block Copolymers Complexation with DNA
Emi Haladjova, et al.
The Journal of Physical Chemistry B, 120, 2586-2595 (2016)
Jeffrey M Ting et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-04)
A series of model polyelectrolyte complex micelles (PCMs) was prepared to investigate the consequences of neutral and zwitterionic chemistries and distinct charged cores on the size and stability of nanocarriers. Using aqueous reversible addition-fragmentation chain transfer (RAFT) polymerization, we synthesized
Nunzio Cennamo et al.
Sensors (Basel, Switzerland), 18(6) (2018-06-08)
A novel Molecularly Imprinted Polymer (MIP) able to bind perfluorinated compounds, combined with a surface plasmon resonance (SPR) optical fiber platform, is presented. The new MIP receptor has been deposited on a D-shaped plastic optical fiber (POF) covered with a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service