Skip to Content
Merck
All Photos(2)

Key Documents

452556

Sigma-Aldrich

Ethanol-OD

99 atom % D

Synonym(s):

Ethan(ol-d), Ethyl alcohol-OD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OD
CAS Number:
Molecular Weight:
47.07
Beilstein:
1731246
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
technique(s):
NMR: suitable
bp:
78 °C (lit.)

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

expl. lim.

3.5-15 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.5000% water
water

refractive index

n20/D 1.359 (lit.)

bp

78 °C (lit.)

mp

-114 °C (lit.)

density

0.806 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i3D

InChI key

LFQSCWFLJHTTHZ-WFVSFCRTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethan(ol-d) (ethanol-d, ethanol-d1) is a deuterated NMR solvent useful in NMR-based research and analyses. It can be prepared by reacting tetraethylorthosilicate with deuterium oxide. The hydrogen bonding in ethanol-d1 has been investigated based on Raman spectral data. The far infrared (FIR) spectrum of ethanol-d1 has been studied using ab initio calculations.

Application

Ethan(ol-d) may be used as a source of deuterium for the deuterium exchange reaction of ethyl acetoacetate.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ab initio determination of the far infrared spectra of some isotopic varieties of ethanol.
Senent ML, et al.
J. Chem. Phys. , 112(13), 5809-5819 (2000)
Deuterium Exchange in Ethyl Acetoacetate: An Undergraduate GC-MS Experiment.
Heinson CD, et al.
Journal of Chemical Education, 82(5), 787-787 (2005)
A quantitative Raman spectroscopic investigation of hydrogen-bonding in ethanol and ethanol-d1.
Spectrochimica Acta Part A: Molecular Spectroscopy.
Edwards HGM, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 45(11), 1165-1171 (1989)
Synthesis of ethanol-d.
Pasto DJ and Meyer GR.
The Journal of Organic Chemistry, 33(3), 1257-1257 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service