Skip to Content
Merck
All Photos(1)

Key Documents

452378

Sigma-Aldrich

Tellurium

shot, ≤2 mm, 99.999% trace metals basis

Synonym(s):

Tellurium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Te
CAS Number:
Molecular Weight:
127.60
EC Number:
MDL number:
UNSPSC Code:
12141913
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

shot

resistivity

5.8-33 μΩ-cm, 20°C

particle size

≤2 mm

bp

990 °C (lit.)

mp

450 °C (lit.)

density

6.24 g/mL at 25 °C (lit.)

SMILES string

[Te]

InChI

1S/Te

InChI key

PORWMNRCUJJQNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains low oxide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 4 - Lact. - Repr. 1B - Skin Sens. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Guizhong Wang et al.
Optics express, 21(4), 4703-4708 (2013-03-14)
The octave-spanning spectrum was generated in a tellurite glass based microstructured fiber pumped by a 528 MHz repetition rate Yb:fiber ring laser without amplification. The laser achieved 40% output optical-to-optical efficiency with the output power of 410 mW. By adjusting
Biomethylation of selenium and tellurium: microorganisms and plants.
Thomas G Chasteen et al.
Chemical reviews, 103(1), 1-25 (2003-01-09)
Raymond J Turner et al.
Biotechnology advances, 30(5), 954-963 (2011-09-13)
Here, we overview the most recent advances in understanding the bacterial mechanisms that stay behind the reduction of tellurium oxyanions in both planktonic cells and biofilms. This is a topic of interest for basic and applied research because microorganisms are
Zhe Liu et al.
Optics express, 21(6), 7799-7810 (2013-04-03)
ZnTe is an important p-type semiconductor with great applications as field-effect transistors and photodetectors. In this paper, individual ZnTe nanowires based field-effect transistors was fabricated, showing evident p-type conductivity with an effect mobility of 11.3 cm(2)/Vs. Single ZnTe nanowire based
Hridyesh Kumar et al.
Carbohydrate polymers, 97(2), 327-334 (2013-08-06)
The present work describes synthesis of water soluble L-cysteine-functionalized CdTe quantum dots (QDs) with size tunable emission at different time intervals for chitosan based film. The characterization of the synthesized CdTe QDs-chitosan film was made by Fourier transform infrared (FTIR)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service