Skip to Content
Merck
All Photos(2)

Documents

450901

Sigma-Aldrich

Holmium(III) chloride

anhydrous, powder, 99.9% trace metals basis

Synonym(s):

Holmium chloride, Holmium(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HoCl3
CAS Number:
Molecular Weight:
271.29
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: holmium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

718 °C (lit.)

SMILES string

Cl[Ho](Cl)Cl

InChI

1S/3ClH.Ho/h3*1H;/q;;;+3/p-3

InChI key

PYOOBRULIYNHJR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B C Satishkumar et al.
Journal of nanoscience and nanotechnology, 3(1-2), 159-163 (2003-08-12)
Nanowires of magnetic metals (Fe, Co, Ho, Gd) have been synthesized inside the hollow interiors of single-wall carbon nanotubes (SWNTs) by filling SWNTs with precursor metal chlorides and subsequent reduction. SWNTs have been filled by either the melt-phase sealed-tube reaction
L Mircevová et al.
Medical biology, 62(6), 326-330 (1984-01-01)
Phagocytosis of polymorphonuclear leucocytes treated with NaF, HoCl3 and adenosine were studied. The highest concentration used was 25 mM of NaF, 25 mM of adenosine and 5 mM of HoCl3. It was ascertained that these substances, inhibitors of erythrocyte contractile
Takefumi Yoshida et al.
Dalton transactions (Cambridge, England : 2003), 48(21), 7144-7149 (2018-09-29)
Heterometallic Ln-Pt complexes, with the formula [Ln2Pt3(H2O)2(SAc)12] (Ln = Ho(1), Er(2); SAc = thioacetate), were synthesized. From natural bond orbital (NBO) and local orbital locator (LOL) analyses and X-ray absorption fine structure (XAFS) measurements, it was clear that the Ln-Pt
Olga Matsarskaia et al.
The journal of physical chemistry. B, 123(9), 1913-1919 (2019-02-01)
Liquid-liquid phase separation (LLPS) in protein systems is relevant for many phenomena, from protein condensation diseases to subcellular organization to possible pathways toward protein crystallization. Understanding and controlling LLPS in proteins is therefore highly relevant for various areas of (biological)
Daqin Chen et al.
The journal of physical chemistry letters, 6(14), 2833-2840 (2015-08-13)
A strategy to achieve 808 nm excited single-band red upconversion luminescence of Ho(3+) via the core-shell nanoarchitecture design was provided. Specifically, the synthesized Yb/Ho/Ce: NaGdF4@Yb/Nd: NaYF4 active-core@active-shell nanoparticles were evidenced to enable high-content doping of Nd(3+) (∼10 mol %) in

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service