Skip to Content
Merck
All Photos(3)

Key Documents

450456

Sigma-Aldrich

4,4′-(1-Phenylethylidene)bisphenol

99%

Synonym(s):

Bisphenol AP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(C6H5)(C6H4OH)2
CAS Number:
Molecular Weight:
290.36
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

188-191 °C (lit.)

SMILES string

CC(c1ccccc1)(c2ccc(O)cc2)c3ccc(O)cc3

InChI

1S/C20H18O2/c1-20(15-5-3-2-4-6-15,16-7-11-18(21)12-8-16)17-9-13-19(22)14-10-17/h2-14,21-22H,1H3

InChI key

VOWWYDCFAISREI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Dipa Lalwani et al.
Ecotoxicology and environmental safety, 200, 110718-110718 (2020-05-29)
Bisphenol A (BPA) has been frequently found in surface waters worldwide, and its estrogenic effects in humans are well documented. Nevertheless, less is known about other bisphenol analogues (BPs), such as bisphenol S (BPS) and bisphenol F (BPF) which are
Chien-Shu Chen et al.
Journal of medicinal chemistry, 52(9), 2716-2723 (2009-05-08)
Hepatitis C virus nonstructural protein 3 (HCV NS3) helicase is believed to be essential for viral replication and has become an attractive target for the development of antiviral drugs. A fluorescence resonant energy transfer helicase assay was established for fast
Marie-Katherin Zühlke et al.
Applied microbiology and biotechnology, 104(8), 3569-3583 (2020-03-04)
Comparative analyses determined the relationship between the structure of bisphenol A (BPA) as well as of seven bisphenol analogues (bisphenol B (BPB), bisphenol C (BPC), bisphenol E (BPE), bisphenol F (BPF), bisphenol Z (BPZ), bisphenol AP (BPAP), bisphenol PH (BPPH))
Yi Chen et al.
Environmental pollution (Barking, Essex : 1987), 238, 299-305 (2018-03-25)
Bisphenols and triclosan (TCS) are widely used in consumer products. However, knowledge on human exposure to these anthropogenic chemicals has remained limited in China, especially for children. In this study, concentrations of seven bisphenols and TCS were determined in 283
Na Li et al.
Talanta, 201, 194-203 (2019-05-28)
A novel magnetic nitrogen-doped reduced graphene oxide (Fe3O4@N-RGO) had been fabricated for the first time on the basis of a simple solvothermal method and then was successfully applied to extract four bisphenol endocrine disruptors (bisphenol A, bisphenol B, bisphenol F

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service