447323
2,5-Di-tert-butyl-4-methoxyphenol
97%
Synonym(s):
2,5-Di-tert-butyl-4-hydroxyanisole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3OC6H2[C(CH3)3]2OH
CAS Number:
Molecular Weight:
236.35
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
99-102 °C (lit.)
SMILES string
COc1cc(c(O)cc1C(C)(C)C)C(C)(C)C
InChI
1S/C15H24O2/c1-14(2,3)10-9-13(17-7)11(8-12(10)16)15(4,5)6/h8-9,16H,1-7H3
InChI key
FLLRQABPKFCXSO-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Qu et al.
Biochimica et biophysica acta, 1501(2-3), 99-115 (2000-06-06)
Phosphatidylserine (PS) was exposed at the surface of human umbilical vein endothelial cells (HUVECs) and cultured cell lines by agonists that increase cytosolic Ca(2+), and factors governing the adhesion of T cells to the treated cells were investigated. Thrombin, ionophore
Sensitive high-performance liquid chromatographic method for the routine determination of butylated hydroxyanisole in plasma.
H Verhagen et al.
Journal of chromatography, 413, 282-286 (1987-01-23)
Efficacy of solid acids in the synthesis of butylated hydroxy anisoles by alkylation of 4-methoxyphenol with MTBE.
Yadav GD and Rahuman MSMM.
Applied Catalysis A: General, 1, 113-123 (2003)
T Mizutani
Research communications in chemical pathology and pharmacology, 50(1), 125-133 (1985-10-01)
Dietary administration of butylated hydroxyanisole (BHA) and its analogs, 2-tert-butyl-1,4-dimethoxybenzene, 2,5-di-tert-butyl-4-methoxyphenol, and 2,6-di-tert-butyl-4-methoxyphenol resulted in complete protection against the lung toxicity of butylated hydroxytoluene (BHT) in mice. The protective effects of these compounds could not be accounted for by their
A Iwado et al.
Chemical & pharmaceutical bulletin, 48(11), 1831-1832 (2000-11-22)
Anion-exchange resins modified with metal-porphine (M-Pr) have been investigated to develop a solid catalyst in the oxidative reaction of phenols by O2 in air. Co-Pr, which is easily prepared and separable from the reaction mixture, has been proved to accelerate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service