All Photos(1)
About This Item
Linear Formula:
HN(CH2CO2C2H5)2
CAS Number:
Molecular Weight:
189.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.435 (lit.)
bp
208 °C (lit.)
density
1.056 g/mL at 25 °C (lit.)
functional group
amine
ester
SMILES string
CCOC(=O)CNCC(=O)OCC
InChI
1S/C8H15NO4/c1-3-12-7(10)5-9-6-8(11)13-4-2/h9H,3-6H2,1-2H3
InChI key
LJDNMOCAQVXVKY-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A new route synthesis of immobilized-polysiloxane iminodiacetic acid ligand system, its characterization and applications.
El-Ashgar NM, et al.
Materials Letters, 61(23), 4533-4558 (2007)
Shiu-Chun Chuang et al.
Journal of chromatography. A, 1044(1-2), 229-236 (2004-09-10)
Polystyrene-divinylbenzene copolymer functionalized with copper-iminodiacetate complex was prepared in situ for the capillary electrochromatographic separation of amino acids and oligopeptides. The steps included silanization of the fused-silica capillary column, functionalization, polymerization, hydrolysis and complexation. For having a homogeneous polymerization system
M Colvin et al.
Cancer research, 36(3), 1121-1126 (1976-03-01)
The relative alkylating activities of two of the cytotoxic metabolites of cyclophosphamide, phosphoramide mustard and nornitrogen mustard, have been studied at pH 4.6 and 7.4. The products formed on alkylation of ethanethiol by these metabolites have been identified, confirming that
Zhenyu Gao et al.
Journal of biomaterials science. Polymer edition, 28(10-12), 1036-1050 (2017-01-31)
As novel magnetic resonance imaging (MRI) contrast agent, gadofullerene encapsulated redox nanoparticles (Gd
Highly effective fluorescent and colorimetric sensors for pyrophosphate over H2PO4-in 100% aqueous solution.
Jang YJ, et al.
The Journal of Organic Chemistry, 70(23), 9603-9606 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service