Skip to Content
Merck
All Photos(3)

Key Documents

442909

Sigma-Aldrich

Calcium chloride hexahydrate

≥95%

Synonym(s):

Calcium dichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 6H2O
CAS Number:
Molecular Weight:
219.08
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
solubility:
219.1 g/L at 20 °C

grade

for analytical purposes

Assay

≥95%

form

solid

mp

30 °C

solubility

219.1 g/L at 20 °C

density

1.71 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.6H2O/h;2*1H;6*1H2/q+2;;;;;;;;/p-2

InChI key

QHFQAJHNDKBRBO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium chloride hexahydrate is a non-toxic salt hydrate that can be used in phase change heat storage of low temperature heat. It has a latent heat of fusion as high as 170-190 kJ/Kg and a melting temperature of 29-30°C.

Application

Calcium chloride hexahydrate is a phase changing material (PCM) that is widely used in solar energy storage and building applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase change material (PCM) based thermal management system for cool energy storage application in building: An experimental study
Tyagi VV, et al.
Energy and Buildings, 51(5), 248-254 (2012)
Experimental investigations on heat transfer in phase change materials (PCMs) embedded in porous materials
Zhou D and Zhao C
Applied Thermal Engineering, 31(5), 970-977 (2011)
Salt hydrates as latent heat storage materials: Thermophysical properties and costs
Kenisarin M and Mahkamov K
Solar Energy Materials and Solar Cells, 145(5), 255-286 (2016)
Phase change behaviour of some latent heat storage media based on calcium chloride hexahydrate
Carlsson Bo
Solar Energy, 83(4), 485-500 (2009)
A novel calcium chloride hexahydrate-based deep eutectic solvent as a phase change materials
Shahbaz K, et al.
Solar Energy Materials and Solar Cells, 155(4), 147-154 (2016)

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service