Skip to Content
Merck
All Photos(1)

Key Documents

440302

Sigma-Aldrich

[3-(2-Aminoethylamino)propyl]trimethoxysilane

technical grade, ≥80%

Synonym(s):

Dow Corning® product Z-6020, N-(2-Aminoethyl)-3-(trimethoxysilyl)propylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3Si(CH2)3NHCH2CH2NH2
CAS Number:
Molecular Weight:
222.36
Beilstein:
636230
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

≥80%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

146 °C/15 mmHg (lit.)

density

1.028 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CO[Si](CCCNCCN)(OC)OC

InChI

1S/C8H22N2O3Si/c1-11-14(12-2,13-3)8-4-6-10-7-5-9/h10H,4-9H2,1-3H3

InChI key

PHQOGHDTIVQXHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • [3-(2-Aminoethylamino)propyl]trimethoxysilane has been used in the synthesis of various polyhedral silsesquioxanes, (RSiO1.5)n (SSQO).
  • It can be functionalized onto the mesoporous SBA-15 silica to be used as matrices for controlled drug delivery.
  • It is also used in the synthesis of silane-based polymer, which is used in conducting instant adhesives.

Legal Information

Repackaged by Aldrich Chemical Company, Inc. Manufactured by Dow Corning Corporation, U.S.A.
Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Sens. 1B - STOT RE 2 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imprinted polysilsesquioxanes for the enhanced recognition of metal ions.
Burleigh M C, et al.
Chemistry of Materials, 13(8), 2537-2546 (2001)
Conducting instant adhesives by grafting of silane polymer onto expanded graphite.
Mondal T, et al.
ACS Applied Materials & Interfaces, 6(18), 16097-16105 (2014)
Synthesis and characterization of polyhedral silsesquioxanes bearing bulky functionalized substituents.
Fasce D P, et al.
Macromolecules, 32(15), 4757-4763 (1999)
One-step synthesis of polyhedral silsesquioxanes bearing bulky substituents: UV-MALDI-TOF and ESI-TOF mass spectrometry characterization of reaction products.
Fasce D P, et al.
Macromolecules, 34(11), 3534-3539 (2001)
Surface-functionalization of mesoporous SBA-15 silica materials for controlled release of methylprednisolone sodium hemisuccinate: Influence of functionality type and strategies of incorporation.
Ortiz-Bustos J, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 240, 236-245 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service