Skip to Content
Merck
All Photos(1)

Key Documents

435694

Sigma-Aldrich

Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride solution

42 wt. % in methanol

Synonym(s):

Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3O)3Si(CH2)3N(CH3)2(CH2)17CH3]Cl
CAS Number:
Molecular Weight:
496.28
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

concentration

42 wt. % in methanol

impurities

8 wt. % (3-chloropropyl)trimethoxysilane

refractive index

n20/D 1.3889

density

0.88 g/mL at 25 °C

SMILES string

[Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC

InChI

1S/C26H58NO3Si.ClH/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-24-27(2,3)25-23-26-31(28-4,29-5)30-6;/h7-26H2,1-6H3;1H/q+1;/p-1

InChI key

WSFMFXQNYPNYGG-UHFFFAOYSA-M

General description

Dimethyloctadecy[3-(trimethoxysilyl)propyl]ammonium chloride solution (DTSACl) is a silane coupling reagent.
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride (DTSACl) is an organosilane which can be used as a silica precursor. It forms an oil soluble solution that can act as an emulsion stabilizer.

Application

DTSACl functionalized silica nanoparticles incorporated in thermochromic dyes can be used in textile industries. They are also used as quaternary ammonium groups for the modification of nanocelluose/graphene oxide based membranes to be used as stable solid state electrolytes for zinc air batteries.
DTSACl may be used for modification of silica.{61} It was used to functionalize Fe3O4/silica core-shell nanoparticles. {62} DTSACl may be used as a precursor to form silica, 3 and MeMFI zeolite.{63} It acts as a mesopore-generating agent. {64}

Disclaimer

This product is not to be used as biocide.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laminated Cross-Linked Nanocellulose/Graphene Oxide Electrolyte for Flexible Rechargeable Zinc-Air Batteries.
Zhang J, et al.
Advanced Energy Materials, 6(14), 1600476-1600476 (2016)
Silica nanoparticles functionalized with a thermochromic dye for textile applications.
Ribeiro LS, et al.
J. Mater. Sci., 48(14), 5085-5092 (2013)
Formation of oil filled nanocapsules with silica shells modified by sequential adsorption of polyelectrolytes.
Szczepanowicz K, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 441, 885-889 (2014)
Mingqiang Wang et al.
Science robotics, 5(45) (2020-10-07)
Batteries with conformal shape and multiple functionalities could provide new degrees of freedom in the design of robotic devices. For example, the ability to provide both load bearing and energy storage can increase the payload and extend the operational range
Biodiesel production by transesterification using tetraalkylammonium hydroxides immobilized onto SBA-15 as a solid catalyst.
Xie W and Fan M
Chemical Engineering Journal, 239, 60-67 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service