Skip to Content
Merck
All Photos(3)

Documents

433098

Sigma-Aldrich

5-Bromo-1-indanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7BrO
CAS Number:
Molecular Weight:
211.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

126-129 °C (lit.)

SMILES string

Brc1ccc2C(=O)CCc2c1

InChI

1S/C9H7BrO/c10-7-2-3-8-6(5-7)1-4-9(8)11/h2-3,5H,1,4H2

InChI key

KSONICAHAPRCMV-UHFFFAOYSA-N

General description

5-Bromo-1-indanone is a 1-indanone derivative. Its physical properties like density, freezing point and refractive index have been determined. It participates in the synthesis of the imidazolyl and triazolyl substituted biphenyl compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Zhuang et al.
Bioorganic & medicinal chemistry, 8(6), 1245-1252 (2000-07-15)
The synthesis of a new series of P450 17 inhibitors is described. The imidazol-1-yl compounds 5 showed strong inhibition of P450 17 rat and especially human enzyme, the most active compounds being 5ax, 5ay and 5bx with IC50 values of
Turn-On Fluorogenic Probes for the Selective and Quantitative Detection of the Cyanide Anion from Natural Sources.
Gomez T, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 8(6), 1271-1278 (2013)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 229-229 (2015)
Andrew K Takle et al.
Bioorganic & medicinal chemistry letters, 16(2), 378-381 (2005-11-02)
A novel triarylimidazole derivative, SB-590885 (33), bearing a 2,3-dihydro-1H-inden-1-one oxime substituent has been identified as a potent and extremely selective inhibitor of B-Raf kinase.
Brian E Fink et al.
Bioorganic & medicinal chemistry letters, 16(6), 1532-1536 (2006-01-03)
A novel series of 17beta-hydroxysteroid dehydrogenase type 3 (17beta-HSD3) inhibitors has been identified. These inhibitors, based on a dibenzazocine core, exhibited picomolar to low nanomolar inhibition of 17beta-HSD3 in cell-free enzymatic as well as in cell-based transcriptional reporter assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service