Skip to Content
Merck
All Photos(2)

Key Documents

430218

Sigma-Aldrich

Poly[4,4′-methylenebis(phenyl isocyanate)-alt-1,4-butanediol/di(propylene glycol)/polycaprolactone]

pellets, MDI-polyester/polyether polyurethane.

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

abrasion resistance

10 mg (Taber, H-22 wheel)

form

beads
pellets

melt index

4 g/10 min (190°C/8.7kg)

hardness

84 (Shore A, ASTM D 2240)

transition temp

Tg −40 °C (DSC)
softening point 85 °C (Vicat, ASTM D 1525)

density

1.18 g/mL at 25 °C (lit.)

SMILES string

N(c1ccc(cc1)Cc2ccc(cc2)NC(=O)O)C(=O)O.OCCCCCC(=O)O.OCCCCO.OCCCO

InChI key

UTAMRXGORNGYAA-UHFFFAOYSA-N

General description

Poly[4,4′-methylenebis(phenyl isocyanate)-alt-1,4-butanediol/di(propylene glycol)/polycaprolactone] (PU/PCL) is a polyurethane solution that can be used as a shape memory polymer and a reinforcing unit for enhancing the mechanical and thermal properties of the base materials. It is biocompatible and finds usage in endothelial proliferations. PU/PCL contains polycaprolactone (PCL) that facilitates the hydrophobic properties of the polymeric matrix.

Application

Automotive fascia, cable jackets, side trim, fabric laminates, health care tubing, catheters, transdermal patches and in seals casters, shoes, bushings and wheels.
PU/PCL can be used in the formation of a polyurethane membrane for effective gas separation. It can also be used to synthesize reinforced hydrogel with good thermo-mechanical properties for biomedical applications.

Features and Benefits

Tough, high-performance, wear-resistant elastomer with good clarity, chemical resistance, flexibility and toughness. Can be used in current form or further polymerized. Low compression set

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuo-jun Hu et al.
BMC cardiovascular disorders, 12, 115-115 (2012-12-05)
In a previous in vitro study, we confirmed that small-caliber nanofibrous polyurethane (PU) vascular grafts have favorable mechanical properties and biocompatibility. In the present study, we examined the in vivo biocompatibility and stability of these grafts. Forty-eight adult male beagle
The in vivo performance of small-caliber nanofibrous polyurethane vascular grafts
Hu Z, et al.
BMC Cardiovascular Disorders, 12(1), 115-115 (2012)
Strong fiber-reinforced hydrogel
Agrawal A, et al.
Acta Biomaterialia, 9(2), 5313-5318 (2013)
A hybrid electrospun PU/PCL scaffold satisfied the requirements of blood vessel prosthesis in terms of mechanical properties, pore size, and biocompatibility
Nguyen T, et al.
Journal of Biomaterials Science. Polymer Edition, 24(14), 1692-1706 (2013)
Mélusine Bouchet et al.
Materials science & engineering. C, Materials for biological applications, 100, 715-723 (2019-04-06)
The search for novel, more compliant vascular grafts for the replacement of blood vessels is ongoing, and predictive tools are needed to identify the most promising biomaterials. A simple analytical model was designed that enables the calculation of the ratio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service