Skip to Content
Merck
All Photos(1)

Documents

424285

Sigma-Aldrich

3-Octylthiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20S
CAS Number:
Molecular Weight:
196.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

106-107 °C/3 mmHg (lit.)

density

0.92 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCc1ccsc1

InChI

1S/C12H20S/c1-2-3-4-5-6-7-8-12-9-10-13-11-12/h9-11H,2-8H2,1H3

InChI key

WQYWXQCOYRZFAV-UHFFFAOYSA-N

General description

3-Octylthiophene is an alkyl thiophene derivative. It has been synthesized by the reaction of 3-bromothiophene with octylmagnesium bromide.

Application

3-Octylthiophene may be used as a starting material in the synthesis of the following:
  • 2,5-dibromo-3-octylthiophene
  • poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene)
  • regioregular poly(3-octylthiophene) (POT)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Crystalline diblock conjugated copolymers: Synthesis, self-assembly, and microphase separation of poly (3-butylthiophene)-b-poly (3-octylthiophene).
Wu PT, et al.
Macromolecules, 42(7), 2317-2320 (2009)
Soluble Phenanthrenyl-Imidazole-Presenting Regioregular Poly (3-octylthiophene) Copolymers Having Tunable Bandgaps for Solar Cell Applications.
Chang YT, et al.
Advances in Functional Materials, 17(16), 3326-3331 (2007)
New convenient synthesis of highly regioregular poly (3-octylthiophene) based on the Suzuki coupling reaction.
Guillerez S and Bidan G.
Synthetic Metals, 93(2), 123-126 (1998)
Said A Hassan et al.
Mikrochimica acta, 187(9), 491-491 (2020-08-09)
The major objective of this work was to develop a portable, disposable, cost-effective, and reliable POC solid-state electrochemical sensor based on potentiometric transduction to detect benzodiazepine abuse, mainly diazepam (DZP), in biological fluids. To achieve that, microfabricated Cu electrodes on
Nannan Jian et al.
Physical chemistry chemical physics : PCCP, 21(13), 7174-7182 (2019-03-20)
Conjugated fluorophores have been extensively used for fluorescence sensing of various substances in the field of life processes and environmental science, due to their noninvasiveness, sensitivity, simplicity and rapidity. Most existing conjugated fluorophores exhibit excellent light-emitting performance in dilute solutions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service