Skip to Content
Merck
All Photos(2)

Key Documents

420247

Sigma-Aldrich

(+)-Borneol

97%

Synonym(s):

endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2038056
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

optical activity

[α]20/D +36°, c = 5 in ethanol

mp

206-209 °C (lit.)

functional group

hydroxyl

SMILES string

[H][C@@]12CC[C@@](C)([C@@H](O)C1)C2(C)C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10+/m1/s1

InChI key

DTGKSKDOIYIVQL-WEDXCCLWSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Borneol, a bicyclic monoterpene, is found in the essential oils of medicinal plants and is commonly used in traditional Chinese medicine for analgesia and anaesthesia.[1]

Application

(+)-Borneol has been used to study its antiapoptotic, antioxidative and neuroprotective effect in human neuroblastoma cells (SH-SY5Y).[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Borneol alleviates oxidative stress via upregulation of Nrf2 and Bcl-2 in SH-SY5Y cells.
Hur J, et al.
Pharmaceutical Analysis, 51(1), 30-35 (2013)
(+)-And (-)-borneol: efficacious positive modulators of GABA action at human recombinant a 1 ? 2 ? 2L GABA A receptors.
Granger RE, et al.
Biochemical Pharmacology, 69(7), 1101-1111 (2005)
Jianyu Su et al.
Journal of food science, 77(6), C658-C664 (2012-05-16)
The aims of this study were to optimize the preparation conditions of natural borneol/β-cyclodextrin (NB/β-CD) inclusion complex by ultrasound method, and to investigate its improvement of stability and solubility. The complex was characterized by different various spectroscopic techniques including Fourier
Jing-yi Zhao et al.
Journal of Zhejiang University. Science. B, 13(12), 990-996 (2012-12-12)
Borneol, a monoterpenoid alcohol, is used widely, particularly in combined formulas for preventing and curing cardiovascular and cerebrovascular diseases in traditional Chinese medicine. In order to understand the blood and brain pharmacokinetics after intravenous, intranasal, or oral administration and to
Xiao Song et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 36(18), 2489-2492 (2012-01-20)
To investigate the absorption characteristic of borneol. Using single pass perfusion model, the active ingredient of borneol were detected by GC. The drug concentration, perfusion rate and pH value on the absorption of borneol were studied. Perfusion rate on the

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service