Skip to Content
Merck
All Photos(1)

Key Documents

409332

Sigma-Aldrich

Cobalt(II) chloride

AnhydroBeads, −10 mesh, 99.995% trace metals basis

Synonym(s):

Cobalt dichloride, Cobalt muriate, Cobaltous chloride, Dichlorocobalt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2
CAS Number:
Molecular Weight:
129.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for synthesis
synthesis grade
form:
beads
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

vapor pressure

40 mmHg ( 0 °C)

product line

AnhydroBeads

Assay

99.995% trace metals basis

form

beads

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

724 °C (lit.)

SMILES string

Cl[Co]Cl

InChI

1S/2ClH.Co/h2*1H;/q;;+2/p-2

InChI key

GVPFVAHMJGGAJG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Precursor for rive-coordinate, heteroleptic thoilate/N complexes which are petential models for cysteine-ligated metalloenzyme activity.[1]
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoner, S.C. et al.
Inorganic Chemistry, 37, 5721-5721 (1998)
Cobalt (II) chloride catalysed coupling of thiols and anhydrides: a new and efficient synthesis of thiol esters.
Ahmad S and Iqbal J.
Tetrahedron Letters, 27(32), 3791-3794 (1986)
Ru-Pan Wang et al.
The journal of physical chemistry. C, Nanomaterials and interfaces, 121(45), 24919-24928 (2017-11-25)
We show that with 2p3d resonant inelastic X-ray scattering (RIXS) we can accurately determine the charge-transfer parameters of CoF
Cobalt (II) chloride catalyzed acylation of alcohols with acetic anhydride: scope and mechanism.
Iqbal J and Srivastava RR.
The Journal of Organic Chemistry, 57(7), 2001-2007 (1997)
An unusual chemoselectivity in cobalt (II) chloride catalysed cleavage of oxiranes with anilines: A highly regioselective synthesis of β-amino alcohols.
Iqbal J and Pandey A.
Tetrahedron Letters, 31(4), 575-576 (1990)

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service