Skip to Content
Merck
All Photos(2)

Key Documents

404454

Sigma-Aldrich

(S,S)-(+)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride

Synonym(s):

(S,S)-Jacobsen’s catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[[(CH3)3C]2C6H2-2-(O-)CH=N]2C6H10]MnCl
CAS Number:
Molecular Weight:
635.20
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

mp

330-332 °C (lit.)

SMILES string

CC(C)(C)c1cc2C=[N@@H]3[C@H]4CCCC[C@@H]4[N@H]5=Cc6cc(cc(c6O[Mn]35(Cl)Oc2c(c1)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C36H54N2O2.ClH.Mn/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12;;/h17-22,29-30,39-40H,13-16H2,1-12H3;1H;/q;;+3/p-3/b37-21+,38-22+;;/t29-,30-;;/m0../s1

InChI key

LJVAWOSDJSQANR-RUIQGICGSA-K

Looking for similar products? Visit Product Comparison Guide

General description

(S,S)-(+)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride is a metal-salen complex that can catalyze a variety of asymmetric transformations.

Application

Reactant involved in:
  • Studies of intercalative binding with ct-DNA and radical scavenging antioxidant activity
  • Immobilization on phenoxy-modified zirconium poly(styrenephenylvinylphosphonate)
  • Synthesis of derivatvies for studies of conformational models of enantioselective reactions

Catalyst involved in:
  • Asymmetric epoxidation of olefins
  • Immobilization on ionic liquid modified mesoporous silica for oxidative kinetic resolution of racemic secondary alcohols†
  • Immobilization on diamine modified zirconium olig-styrenyl phosphonate hydrogen phosphate for epoxidation of styrene
Salen (Mn)-catalyzed asymmetric epoxidation of unfunctionalized olefins. High enantioselectivities and yields are obtained for a variety of substrates, especially cis-alkenes. A few applications include the synthesis of the taxol side chain and cis-1-amino-2-indanol. Jacobsen′s catalyst has also been used in the asymmetric α-hydroxylation of silyl enol ethers. Catalyst for the enantioselective epoxidation of a variety of olefins.

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Senanayake, C.H. et al.
Tetrahedron Letters, 37, 3271-3271 (1996)
Senanayake, C.H. et al.
Tetrahedron Letters, 36, 3993-3993 (1995)
Adam, W. et al.
Tetrahedron, 37, 6531-6531 (1996)
Zhang, W. Jacobsen, E.N.
The Journal of Organic Chemistry, 56, 2296-2296 (1991)
Lee, N.H. et al.
Tetrahedron Letters, 32, 5055-5055 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service