Skip to Content
Merck
All Photos(1)

Documents

399396

Sigma-Aldrich

Benzoin

purified by sublimation, ≥99.5%

Synonym(s):

α-Hydroxy-α-phenylacetophenone, α-Hydroxybenzyl phenyl ketone, 2-Hydroxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COC6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
391839
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.5%

form

powder

purified by

sublimation

bp

194 °C/12 mmHg (lit.)

mp

134-138 °C (lit.)

SMILES string

OC(c1ccccc1)C(=O)c2ccccc2

InChI

1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H

InChI key

ISAOCJYIOMOJEB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sarvesh Kumar et al.
Archiv der Pharmazie, 345(5), 368-377 (2011-12-23)
The interaction between leukocytes and the vascular endothelial cells (EC) via cellular adhesion molecules plays an important role in the pathogenesis of various inflammatory and autoimmune diseases. Small molecules that block these interactions have been targeted as potential therapeutic agents
Jun He et al.
Electrophoresis, 32(10), 1164-1175 (2011-04-19)
In the present work we report, for the first time, the successful on-line coupling of chiral MEKC (CMEKC) to atmospheric pressure photoionization MS (APPI-MS). Four structurally similar neutral test solutes (e.g. benzoin (BNZ) derivatives) were successfully ionized by APPI-MS. The
Total synthesis and absolute stereochemistry of seragakinone A.
Akiomi Takada et al.
Angewandte Chemie (International ed. in English), 50(10), 2297-2301 (2011-02-26)
Yankai Liu et al.
Organic letters, 14(5), 1310-1313 (2012-03-03)
Expanding upon the recently developed aminocatalytic asymmetric indole-2,3-quinodimethane strategy, a straightforward synthesis of structurally and stereochemically complex tetrahydrocarbazoles has been devised. The chemistry's complexity-generating power was further harnessed by designing a multicatalytic, one-pot Diels-Alder/benzoin reaction sequence to stereoselectively access trans-fused
Direct copper-catalyzed α-arylation of benzyl phenyl ketones with aryl iodides: route towards tamoxifen.
Grégory Danoun et al.
Angewandte Chemie (International ed. in English), 51(51), 12815-12819 (2012-11-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service