Skip to Content
Merck
All Photos(1)

Documents

397326

Sigma-Aldrich

Tetrakis(diethylamido)tin(IV)

Synonym(s):

Tetrakis(diethylamino)tin(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C2H5)2N]4Sn
CAS Number:
Molecular Weight:
407.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

reaction suitability

core: tin

refractive index

n20/D 1.49 (lit.)

bp

110 °C/0.5 mmHg (lit.)

density

1.125 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)[Sn](N(CC)CC)(N(CC)CC)N(CC)CC

InChI

1S/4C4H10N.Sn/c4*1-3-5-4-2;/h4*3-4H2,1-2H3;/q4*-1;+4

InChI key

IFVSPCQTOMZHOP-UHFFFAOYSA-N

General description

Tetrakis(diethylamido)tin(IV) is a volatile compound with four exchangeable ligands. It is widely used as a precursor to produce metal oxide thin films by the chemical vapor deposition method.

Application

Tetrakis(diethylamido)tin(IV) can be used:
  • As a precursor to synthesize mesoporous tin silicate xerogels via non-hydrolytic templated sol–gel synthesis. These xerogels are potential catalysts for aminolysis and for the Meerwein–Ponndorf–Verley reduction.
  • To produce stable metal oxide thin films on the surface of carbon electrode materials.
  • To fabricate an ultrathin layer of SnO2 on carbon nanotubes, which can be used as anodes for lithium-ion batteries. This ultrathin layer helps to enhance the cycling stability and rate capability of anodes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous SnO2-SiO2 and Sn-silica-carbon nanocomposites by novel non-hydrolytic templated sol-gel synthesis
David Skoda, et al.
Royal Society of Chemistry Advances, 6, 68739-68747 (2016)
Electrochemical properties of metal-oxide-coated carbon electrodes prepared by atomic layer deposition
James A Loussaert, et al.
Langmuir, 30, 13707-13715 (2014)
Growth of ultrathin SnO2 on carbon nanotubes by atomic layer deposition and their application in lithium-ion battery anodes
Shengyun Zhu, et al.
Applied Surface Science, 484, 600-609 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service