Skip to Content
Merck
All Photos(2)

Key Documents

394440

Sigma-Aldrich

Hydrazine monohydrobromide

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NNH2 · HBr
CAS Number:
Molecular Weight:
112.96
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

solid

mp

87-92 °C (lit.)

SMILES string

Br.NN

InChI

1S/BrH.H4N2/c;1-2/h1H;1-2H2

InChI key

DVHXJLRODLTJOD-UHFFFAOYSA-N

General description

Hydrazine monohydrobromide (hydrazinium bromide) is a hydrobromide salt of hydrazine. It has been prepared by adding aqueous hydrobromic acid to aqueous hydrazine. Its crystal structure has been investigated. It participates in the synthesis of 2-(2′-furoyl)-4(5)-(2′-furanyl)-1H-imidazole (FFI). It is formed as an intermediate during the formation of phosphaminimides from hydrazide derivatives. The fumes from the pyrolysis of hydrazine monohydrobromide soldering flux have been reported to cause dermatitis.

Application

Hydrazine monohydrobromide (hydrazinium bromide) may be used in the synthesis of bromostannates and bromostannites by reacting with tin bromide and also in soldering flux.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protection of workers against dermatitis on exposure to hydrazine hydrobromide soldering flux.
Cook WA
International Archives of Occupational and Environmental Health, 13(6), 616-618 (1955)
A Facile Synthesis of an Advanced Glycosylation Endproduct 2-(2'-furoyl)-4 (5)-(2'-furanyl)-1 H-imidazole.
Rho T, et al
Synthetic Communications, 27(24), 4315-4318 (1997)
504. Hydrazine. Part IV. The bromostannates and bromostannites of hydrazine, dimethyl ketazine, and pentan-3-one hydrazone.
Pugh, W
Journal of the Chemical Society, 2491-2493 (1953)
Hydrazinium bromide as a solvent.
Seward RP
The Journal of Physical Chemistry, 66(6), 1125-1127 (1962)
The crystal structure of hydrazinium bromide, N2H5Br.
Sakurai K and Tomiie Y
Acta Crystallographica, 5(2), 289-289 (1952)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service