Skip to Content
Merck
All Photos(1)

Documents

391425

Sigma-Aldrich

1-Acetylpyrene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12O
CAS Number:
Molecular Weight:
244.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

86-89 °C (lit.)

SMILES string

CC(=O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C18H12O/c1-11(19)15-9-7-14-6-5-12-3-2-4-13-8-10-16(15)18(14)17(12)13/h2-10H,1H3

InChI key

KCIJNJVCFPSUBQ-UHFFFAOYSA-N

General description

1-Acetylpyrene is a pyrene derivative. Its synthesis has been reported. Its phytophysical properties have been studied using absolute fluorescence quantum yield measurement and time-dependent density functional theory (TD-DFT) calculations. Its ability to interact with human cytochromes P450 2A13, 2A6, and 1B1 and enzyme inhibition has been reported. Its function as an environment-sensitive fluorophore has been investigated.

Application

1-Acetylpyrene is suitable for use in a comparative study on the photoinitiating efficiency of pyrene, 1-acetylpyrene and 1-(bromoacetyl)pyrene for copolymerization of styrene with acrylonitrile. It may be used in the following studies:
  • As a starting material in the synthesis of ethynlypyrene. 1-(1-chlorovinyl)pyrene was also isolated during this reaction.
  • As a starting material in the synthesis of substituted pyrene derivatives incorporated heterocyclic and sugar moieties.
  • Synthesis of (E)-pyrene oxime ester conjugates of carboxylic acids.
  • Synthesis of tertiary alcohols based on 1-acetylpyrene.
  • Synthesis of (E)-N-[1-(pyren-1-yl)ethylidene]chrysene-2-amine.
  • Synthesis of 3,3-di(methylsulfanyl)-1-(1-pyrenyl)-2-propen-1-one.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and study of film-forming properties and light sensitivity of 4-acyloxy-3-methoxy (ethoxy) phenylmethylidene-(chrysen-2-yl) amines.
Dikusar EA, et al.
Russ. J. Gen. Chem., 77(2), 278-281 (2007)
Synthesis and fluorescent properties of 5-(1-pyrenylethynyl)-2'-deoxyuridine-containing oligodeoxynucleotides.
Malakhov AD, et al.
Russian J. Bioorg. Chem., 26(1), 34-44 (2000)
Fundamental photoluminescence properties of pyrene carbonyl compounds through absolute fluorescence quantum yield measurement and density functional theory.
Niko Y, et al.
Tetrahedron, 68(31), 6177-6185 (2012)
1-(Bromoacetyl) pyrene, a novel photoinitiator for the copolymerization of styrene and acrylonitrile.
Mishra A and Daswal S.
Coll. Polymer Sci., 285(4), 397-404 (2007)
Tertiary alcohols based on 1-acetylpyrene.
Dikusar EA.
Russ. J. Org. Chem., 43(10), 1492-1494 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service