Skip to Content
Merck
All Photos(2)

Key Documents

389293

Sigma-Aldrich

Poly(vinyl chloride)

average Mw ~43,000, average Mn ~22,000

Synonym(s):

PVC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCl)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mn ~22,000
average Mw ~43,000

inherent viscosity

0.51 dL/g(lit.)

density

1.4 g/mL at 25 °C (lit.)

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl chloride) (PVC) is a widely utilized thermoplastic material with excellent thermo-mechanical properties.

Application

PVC can be used in a variety of viable applications such as: formation of nanobifiller for potential usage in nanoreactors, modification of pyrogenic products, preparation of PVC based aerogels which can be used as high performance thermal insulation materials

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultralight and Flexible Monolithic Polymer Aerogel with Extraordinary Thermal Insulation by A Facile Ambient Process
Li M, et al.
Advanced Materials Interfaces, 6(13), 1900314-1900314 (2019)
Compositional modification of pyrogenic products using CaCO3 and CO2 from the thermolysis of polyvinyl chloride (PVC)
Lee T, et al.
Green Chemistry, 20(7), 1583-1593 (2018)
Fabrication and characterization of polyvinyl chloride/poly(styrene-co-maleic anhydride) intercalated functional nanobifiller-based composite paper
Khan DM, et al.
International Journal of Polymer Analysis and Characterization, 21(3), 228-243 (2016)
Separation and analysis of low molecular weight plasticizers in poly (vinyl chloride) tubes
Wang Q and Storm BK
Polymer Testing, 24(3), 290-300 (2005)
Christopher B Stipe et al.
Applied spectroscopy, 66(11), 1286-1293 (2012-11-14)
Airborne silica dust (quartz) is common in coal mines and represents a respiratory hazard that can lead to silicosis, a potentially fatal lung disease. With an eye toward developing a portable monitoring device for rapid analysis of silica dust, laser-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service