Skip to Content
Merck
All Photos(1)

Key Documents

379824

Sigma-Aldrich

Potassium persulfate

99.99% trace metals basis

Synonym(s):

Potassium peroxodisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S2O8
CAS Number:
Molecular Weight:
270.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
solid
Pricing and availability is not currently available.

vapor density

9.3 (vs air)

Quality Level

Assay

99.99% trace metals basis

form

solid

reaction suitability

reagent type: oxidant

impurities

≤150.0 ppm Trace Metal Analysis

SMILES string

[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/2K.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

USHAGKDGDHPEEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium persulfate is used to initiate the polymerization of styrene to form monodispersed, surfactant-free polystyrene spheres.[1] Potassium persulfate is used as an oxidant to generate 2,2-azinobis(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) radical cation to measure the antioxidant activities of natural compounds.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Monodisperse Polymer Spheres.
Fierke MA
Material Matters , 3.1, 13-13 (2008)
ESR and X-ray Structure Investigations on the Binding and Mechanism of Inhibition of the Native State of Myeloperoxidase with Low Molecular Weight Fragments
Chavali B, et al.
Applied magnetic resonance, 46.8, 853-873 (2015)
Free radical degradation of chitosan with potassium persulfate.
Hsu SC, et al.
Polymer Degradation and Stability, 75(1), 73-83 (2002)
Potassium persulfate promoted catalytic wet oxidation of fulvic acid as a model organic compound in landfill leachate with activated carbon.
Xu XY, et al.
Chemical Engineering Journal, 200, 25-31 (2012)
Toughening polystyrene by core-shell grafting copolymer polybutadiene-graft-polystyrene with potassium persulfate as initiator.
Di Cai G, et al.
Journal of Industrial and Engineering Chemistry (Amsterdam, Netherlands), 19(3), 823-828 (2013)

Articles

An article regarding common FAQs for initiators and stabilizers.

An article regarding common FAQs for initiators and stabilizers.

An article regarding common FAQs for initiators and stabilizers.

An article regarding common FAQs for initiators and stabilizers.

Protocols

Polymeric spheres serve as crystal templates. Synthesis methods yield large quantities.

Polymeric spheres serve as crystal templates. Synthesis methods yield large quantities.

Polymeric spheres serve as crystal templates. Synthesis methods yield large quantities.

Polymeric spheres serve as crystal templates. Synthesis methods yield large quantities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service