Skip to Content
Merck
All Photos(2)

Documents

379816

Sigma-Aldrich

Potassium tetrachloropalladate(II)

99.99% trace metals basis

Synonym(s):

Potassium palladium(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PdCl4
CAS Number:
Molecular Weight:
326.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

solid

impurities

≤150.0 ppm Trace Metal Analysis

mp

105 °C (dec.) (lit.)

density

2.67 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].Cl[Pd--](Cl)(Cl)Cl

InChI

1S/4ClH.2K.Pd/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

LGCKLDWLSVFMGL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

Features and Benefits

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar. Reacts with bis(dithiolates) to metal-bis(dithiolates) with applications in laser Q-switch materials, optical CD recording media, bar code material and superconductivity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naomi Winckelmans et al.
The journal of physical chemistry. C, Nanomaterials and interfaces, 122(25), 13522-13528 (2018-07-10)
Three dimensional (3D) characterization of structural defects in nanoparticles by transmission electron microscopy is far from straightforward. We propose the use of a dose-efficient approach, so-called multimode tomography, during which tilt series of low and high angle annular dark field
M J McLean et al.
Journal of molecular recognition : JMR, 1(3), 138-151 (1988-06-01)
Five different DNA fragments have been treated with a range of conformationally sensitive reagents in an effort to probe structural changes in DNA associated with binding of the bis-intercalating antibiotic echinomycin. For each probe, the intensity and pattern of its
B L Iverson et al.
Nucleic acids research, 15(19), 7823-7830 (1987-10-12)
Reaction of DNA with K2PdCl4 at pH 2.0 followed by a piperidine workup produces specific cleavage at adenine (A) residues. Product analysis revealed the K2PdCl4 reaction involves selective depurination at adenine, affording an excision reaction analogous to the other chemical
A water-based synthesis of octahedral, decahedral, and icosahedral Pd nanocrystals.
Byungkwon Lim et al.
Angewandte Chemie (International ed. in English), 46(48), 9279-9282 (2007-10-31)
V A Tomilets et al.
Farmakologiia i toksikologiia, 44(1), 77-80 (1981-01-01)
A scheme has been devised that makes it possible to assess a specific mechanism of action of any histamine-releasing agent. The scheme suggests a switch off of different stages of histamine secretion--interaction of a histamine liberator with surface cell receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service