Skip to Content
Merck
All Photos(3)

Key Documents

379506

Sigma-Aldrich

Tris(2,3-epoxypropyl) isocyanurate

Synonym(s):

1,3,5-Tris(oxiran-2-ylmethyl)-1,3,5-triazinane-2,4,6-trione, TGIC, Triglycidyl isocyanurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O6
CAS Number:
Molecular Weight:
297.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

SMILES string

O=C1N(CC2CO2)C(=O)N(CC3CO3)C(=O)N1CC4CO4

InChI

1S/C12H15N3O6/c16-10-13(1-7-4-19-7)11(17)15(3-9-6-21-9)12(18)14(10)2-8-5-20-8/h7-9H,1-6H2

InChI key

OUPZKGBUJRBPGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Muta. 1B - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Airborne contact dermatitis due to triglycidylisocyanurate.
A Dooms-Goossens et al.
Contact dermatitis, 21(3), 202-203 (1989-09-01)
Occupational contact allergy to triglycidyl isocyanurate (TGIC, Tepic).
K Nishioka et al.
Contact dermatitis, 19(5), 379-380 (1988-11-01)
J Rubin et al.
Cancer treatment reports, 71(5), 489-492 (1987-05-01)
Two schedules of teroxirone, a triazine triepoxide, were evaluated in a phase I study. Twenty-six patients were treated on 1 day every 5 weeks at doses of 36-2250 mg/m2. At doses greater than or equal to 1500 mg/m2, severe thrombophlebitis
U Erikstam et al.
Contact dermatitis, 44(1), 13-17 (2001-01-13)
Triglycidyl isocyanurate (TGIC) is mainly used in polyester-based powder paints, but also in laminates, insulating varnishes, coatings and adhesives. Several cases of contact allergy to TGIC have been reported during the last 10 years. Contact allergy to TGIC has developed
Martin Roff et al.
The Annals of occupational hygiene, 48(3), 257-265 (2004-04-03)
This paper describes workplace dermal exposure measurements that were carried out by the Health and Safety Laboratory as part of the EU RISKOFDERM project to measure dust contamination. Exposure to dry powder spray paints was measured at five sites on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service