378836
Magnesium fluoride
random crystals, optical grade, ≥99.99% trace metals basis
Synonym(s):
Sellaite
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
MgF2
CAS Number:
Molecular Weight:
62.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
grade:
optical grade
form:
random crystals
Recommended Products
grade
optical grade
Quality Level
Assay
≥99.99% trace metals basis
form
random crystals
impurities
≤100.0 ppm Trace Metal Analysis
density
3.15 g/mL at 25 °C (lit.)
SMILES string
F[Mg]F
InChI
1S/2FH.Mg/h2*1H;/q;;+2/p-2
InChI key
ORUIBWPALBXDOA-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Controlling the rate of degradation of Mg using magnesium fluoride and magnesium fluoride-magnesium phosphate duplex coatings: This study discusses coatings that control the degradation rate of magnesium, which could be crucial in biomedical applications such as bioresorbable implants (K Ravichandran, SN TSN, 2022).
- Electroforming-Free Bipolar Resistive Switching Memory Based on Magnesium Fluoride: Discusses the development of resistive switching memory devices using magnesium fluoride, focusing on their applications in non-volatile memory technology (NC Das et al., 2021).
- Supercontinuum generation for optical coherence tomography using magnesium fluoride photonic crystal fiber: Explores the use of magnesium fluoride in photonic crystal fibers to enhance supercontinuum generation, which is crucial for improving optical coherence tomography (A Ghanbari et al., 2017).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J Preben Morth et al.
Nature, 450(7172), 1043-1049 (2007-12-14)
The Na+,K+-ATPase generates electrochemical gradients for sodium and potassium that are vital to animal cells, exchanging three sodium ions for two potassium ions across the plasma membrane during each cycle of ATP hydrolysis. Here we present the X-ray crystal structure
Martina Thomann et al.
Journal of biomedical materials research. Part A, 93(4), 1609-1619 (2010-01-15)
MgCa0.8 cylinders (2.5 x 25 mm(2)) were coated with a magnesium-fluoride layer and implanted in the marrow cavities of both tibiae of 10 New Zealand White rabbits. The implantation duration was 3 and 6 months. The implants were clinically well
Jonathan Lellouche et al.
Biomaterials, 30(30), 5969-5978 (2009-08-12)
The ability of bacteria to develop antibiotic resistance and colonize abiotic surfaces by forming biofilms is a major cause of medical implant-associated infections and results in prolonged hospitalization periods and patient mortality. This raises the urgent need to develop compounds
Jonathan Lellouche et al.
Nanomedicine : nanotechnology, biology, and medicine, 8(5), 702-711 (2011-09-29)
Antibiotic resistance has prompted the search for new agents that can inhibit bacterial growth. We recently reported on the antimicrobial and antibiofilm activities of nanosized magnesium fluoride (MgF(2)) nanoparticles (NPs) synthesized in ionic liquid using microwave chemistry. In this article
Jonathan Lellouche et al.
International journal of nanomedicine, 7, 1175-1188 (2012-03-16)
The ability of bacteria to colonize catheters is a major cause of infection. In the current study, catheters were surface-modified with MgF(2) nanoparticles (NPs) using a sonochemical synthesis protocol described previously. The one-step synthesis and coating procedure yielded a homogenous
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service