Skip to Content
Merck
All Photos(1)

Key Documents

378372

Sigma-Aldrich

Silicone oil

viscosity 350 cSt (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-Si(CH3)2O-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

vapor density

>1 (vs air)

vapor pressure

<5 mmHg ( 25 °C)
5 mmHg ( 20 °C)

form

viscous liquid

refractive index

n20/D 1.403 (lit.)

viscosity

350 cSt(25 °C)

bp

>140 °C/0.002 mmHg (lit.)

density

0.968 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Silicone oil is a liquid based siloxane that is part of the methyl silicone fluid system. It has a viscosity of 350 cSt with a refractive index of ~ 1.402 and a dielectric strength of ~ 14 kV/mm. It′s surface tension tends to increase with an increase in the viscosity.

Application

Silicone oil can be used for a variety of applications such as: heat transferring medium in chemical and petrochemical industries, a dielectric coolant, a lubricant and antiflatulent agent, protective coatings for building materials, a cosmetic additive.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silicones
Moretto H, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Recent developments and applications of protective silicone coatings: A review of PDMS functional materials
Eduok U, et al.
Progress in Organic Coatings, 111, 124-163 (2017)
Mechanism of Stabilization of Silicone Oil- Water Emulsions Using Hybrid Siloxane Polymers
Mehta SC and Somasundaran P
Langmuir, 24(9), 4558-4563 (2008)
Daniel T Ginat et al.
JAMA otolaryngology-- head & neck surgery, 139(2), 199-201 (2013-02-23)
Silastic implants are popular for cosmetic cheek augmentation. We describe the computed tomographic (CT) and magnetic resonance imaging (MRI) findings in 5 cases (4 patients) with penetration of the medial aspect of Silastic cheek implants through the anterior maxillary sinus
John R Fowler et al.
The Orthopedic clinics of North America, 44(3), 425-431 (2013-07-06)
Radial head fractures without associated bony or ligamentous injury can be safely treated with internal fixation, if possible, or arthroplasty if nonreconstructable. However, nonreconstructable radial head fractures in association with elbow dislocation and/or ligamentous injury in the elbow or forearm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service