376523
3-(2-Bromoethyl)indole
97%
Synonym(s):
1-Bromo-2-(3-indolyl)ethane, 2-(3-Indolyl)ethyl bromide, 3-(2-Bromoethyl)-1H-indole, 3-(Bromoethyl)-1H-indole
About This Item
Recommended Products
Quality Level
Assay
97%
mp
97-99 °C (lit.)
solubility
chloroform: soluble 25 mg/mL, clear, yellow
SMILES string
BrCCc1c[nH]c2ccccc12
InChI
1S/C10H10BrN/c11-6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6H2
InChI key
NTLAICDKHHQUGC-UHFFFAOYSA-N
General description
Application
- β-carboline derivatives
- 6,7-dihydro-12H-indolo[2,3-a] pyridocolinium bromide
- N-(2-(3-indolyl)ethyl)aza-12-crown-4
- N-(2-(3-Indolyl)ethyl)aza-15-crown-5
- N-(2-(3-Indolyl)ethyl)aza-18-crown-6
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service