Skip to Content
Merck
All Photos(1)

Key Documents

375217

Sigma-Aldrich

2-Chloro-4,6-dimethoxy-1,3,5-triazine

97%

Synonym(s):

2,4-Dimethoxy-6-chloro-1,3,5-triazine, 2,4-Dimethoxy-6-chloro-s-triazine, 2-Chloro-4,6-dimethoxy-1,3,5-triazine, 2-Chloro-4,6-dimethoxy-s-triazine, 4,6-Dimethoxy-2-chloro-s-triazine, 6-Chloro-2,4-dimethoxy-s-triazine, CDMT, Chlorodimethoxytriazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6ClN3O2
CAS Number:
Molecular Weight:
175.57
Beilstein:
148988
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

71-74 °C (lit.)

functional group

chloro

SMILES string

COc1nc(Cl)nc(OC)n1

InChI

1S/C5H6ClN3O2/c1-10-4-7-3(6)8-5(9-4)11-2/h1-2H3

InChI key

GPIQOFWTZXXOOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-4,6-dimethoxy-1,3,5-triazine is a stable, yet highly reactive, peptide coupling agent. It is reported as useful coupling reagent for the purification of peptides. Catalytic amide-forming reactions of 2-chloro-4,6-dimethoxy-1,3,5-triazine has been reported. Procedure for the preparation of multikilogram quantities of 2-chloro-4,6-dimethoxy-1,3,5-triazine has been reported.

Application

2-Chloro-4,6-dimethoxy-1,3,5-triazine may be used in the following studies:
  • The specific labeling of streptavidin by the modular method for affinity labeling (MoAL).
  • The preparation of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, via reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine.
  • The preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts, via reaction with various tertiary amines.
  • The synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride, via coupling with N-methylmorpholine in THF.
A stable, yet highly reactive, peptide coupling agent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved procedure for the large scale preparation of 2-chloro-4, 6-dimethoxy-1, 3, 5-triazine.
Cronin JS, et al.
Synthetic Communications, 26(!8), 3491-3494 (1996)
2-Chloro-4, 6-dimethoxy-1, 3, 5-triazine. A new coupling reagent for peptide synthesis.
Kaminski ZJ.
Synthesis, 10, 917-920 (1987)
Munetaka Kunishima et al.
Chemical & pharmaceutical bulletin, 61(8), 882-886 (2013-08-02)
Effect of the basic property of reactants (tertiary amine catalysts, a substrate amine, and acid neutralizers) on catalytic dehydrocondensation between a carboxylic acid and an amine by using 2-chloro-4,6-dimethoxy-1,3,5-triazine (CDMT) was studied. The reaction yield was affected by the acid-base
Munetaka Kunishima et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(49), 15856-15867 (2012-10-13)
The reaction of 2-chloro-4,6-dimethoxy-1,3,5-triazine (CDMT) with various nitrogen-containing compounds, particularly tertiary amines (tert-amines), has been studied for the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts [DMT-Am(s)]. DMT-Ams derived from aliphatic tert-amines exhibited activity for the dehydrocondensation between a carboxylic acid and an amine
Munetaka Kunishima et al.
Chemical & pharmaceutical bulletin, 62(11), 1146-1150 (2014-11-05)
We studied the specific labeling of streptavidin using the modular method for affinity labeling (MoAL) that we developed based on a catalytic amide-forming reaction using 2-chloro-4,6-dimethoxy-1,3,5-triazine (CDMT) and a tertiary amine catalyst. The primary structures of avidin and streptavidin are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service