Skip to Content
Merck
All Photos(1)

Key Documents

374598

Sigma-Aldrich

Cetyltrimethylammonium hydrogensulfate

99%

Synonym(s):

CTAHSO4, Hexadecyltrimethylammonium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(HSO4)(CH3)3
CAS Number:
Molecular Weight:
381.61
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

mp

250-260 °C (dec.) (lit.)

SMILES string

OS([O-])(=O)=O.CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.H2O4S/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;1-5(2,3)4/h5-19H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

UCRJJNVFJGKYQT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cetyltrimethylammonium hydrogensulfate is commonly used as a surfactant for organic transformation in the aqueous medium.[1][2][3]
Some of the applications include:
  • Rhodium(I)-catalyzed asymmetric hydrogenation of (Z)-methyl-α-acetamidocinnamate.[2]
  • Asymmetric palladium-catalyzed alkylation of 1,3-diphenyl-2-propenyl acetate with dimethyl malonate.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micellar effects upon oxidation of organic sulfides by anionic oxidants
Bacaloglu R, et al.
Journal of Physical Organic Chemistry, 5(4), 171-178 (1992)
Influence of different types of amphiphiles on the rhodium (I) complex-catalyzed asymmetric hydrogenation of (Z)-methyl-α-acetamidocinnamate in aqueous medium
Grassert I, et al.
Tetrahedron, 49(30), 6605-6612 (1993)
Catalytic asymmetric alkylation in water in the presence of surfactants
Sinou D, et al.
advanced synthesis and catalysis, 345(3), 357-363 (2003)
K Ranta et al.
Journal of investigational allergology & clinical immunology, 23(4), 226-233 (2013-08-24)
Immunoglobulin E-mediated allergies have doubled in prevalence during recent decades in developed countries.This increase has been attributed, in part, to high hygiene standards, which have reduced exposure to microbes. The capacity of microbes to induce type 1 helper T cell
Lin Wang et al.
Journal of hazardous materials, 244-245, 681-688 (2012-11-28)
Photocatalytic reduction of aqueous Cr(VI) was successfully achieved on nanostructured SnIn(4)S(8). The SnIn(4)S(8) particles with flower-like nanostructure were synthesized via a facile solvothermal method. UV-vis diffuse reflectance spectra (DRS) indicated that the SnIn(4)S(8) particles had strong absorption in visible region

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service