374156
3-Cyanopropyltriethoxysilane
98%
Synonym(s):
4-(Triethoxysilyl)butyronitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5O)3Si(CH2)3CN
CAS Number:
Molecular Weight:
231.36
Beilstein:
1942237
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.417 (lit.)
bp
100-101 °C/6 mmHg (lit.)
density
0.966 g/mL at 25 °C (lit.)
SMILES string
CCO[Si](CCCC#N)(OCC)OCC
InChI
1S/C10H21NO3Si/c1-4-12-15(13-5-2,14-6-3)10-8-7-9-11/h4-8,10H2,1-3H3
InChI key
VGIURMCNTDVGJM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
3-cyanopropyltriethoxysilane (CPTS) is a trialkoxysilane with a reactive cyanoalkyl group attached to the silicon atom.
Application
CPTS is used in the synthesis of titanium based sorbent for solid phase extraction(SPE) of aromatic amines from river water. It finds its application in the development of organically modified silicate films coated on gold electrodes for the detection of ametryn. It may also be used to modify the surface characteristics of biopolymers by using polar functionalities for the analysis of non-steroidal anti-inflammatory drugs(NSAIDs) in river water.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
208.4 °F - closed cup
Flash Point(C)
98 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Surface modification of polyimide film surface by silane coupling reactions for copper metallization.
Inagaki N, et al.
Journal of Adhesion Science and Technology, 15(7), 749-762 (2001)
Synthesis of functionalized silica nanostructure: Unexpected conversion of cyanopropyl group in chloropropyl one during HCl-catalysed hydrolysis of the corresponding triethoxysilane.
Balan M, et al.
Journal of Molecular Structure, 1110, 150-155 (2016)
Catalytic hydrolysis of cellobiose using different acid-functionalised Fe3O4 magnetic nanoparticles.
Avinash P Ingle et al.
IET nanobiotechnology, 14(1), 40-46 (2020-01-15)
The present study demonstrated the preparation of three different acid-functionalised magnetic nanoparticles (MNPs) and evaluation for their catalytic efficacy in hydrolysis of cellobiose. Initially, iron oxide (Fe3O4)MNPs were synthesised, which further modified by applying silica coating (Fe3O4-MNPs@Si) and functionalised with
Exploring electrochemical reactivity toward ametryn of hybrid silicate films with phosphomolybdic acid.
Olivio PH, et al.
Materials Science and Engineering, B, 229, 13-19 (2018)
Reaction of the Si?Cl bond with trialkyl orthoformates preparation of alkoxy-substituted silanes
Herzog U, et al.
Journal of Organometallic Chemistry, 547(1), 133-139 (1997)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service