Skip to Content
Merck
All Photos(2)

Key Documents

362700

Sigma-Aldrich

B-Chlorocatecholborane

97%

Synonym(s):

2-Chloro-1,3,2-benzodioxaborole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BClO2
CAS Number:
Molecular Weight:
154.36
Beilstein:
2093985
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

56-58 °C (lit.)

SMILES string

Clb1oc2ccccc2o1

InChI

1S/C6H4BClO2/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H

InChI key

AZYGEWXDKHFOKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

B-Chlorocatecholborane is a boron reagent and a Lewis acid, known to facilitate the borylative cyclization of alkynes to yield the borylated heterocycles. It is also used in the preparation of lactones, and thiophenes.

Application

B-Chlorocatecholborane can be used:
  • To prepare 2-arachidonoylglycerol by acetal cleavage of cis-arachidonoylbenzylidene glycerol.
  • To prepare metal boryl complexes (Rh and Ir complexes) through oxidative addition.
  • To remove the trityl group in one of the key steps for the synthesis of (−)-dictyostatin.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of (−)-dictyostatin and stereoisomers
Shin Y, et al.
Tetrahedron, 63(35), 8537-8562 (2007)
Synthesis of rhodium and iridium boryl complexes via oxidative addition of haloboranes
Souza FES, et al.
Inorgorganica Chimica Acta, 358(5), 1501-1509 (2005)
Mild acetal cleavage using B-chlorocatecholborane in the synthesis of rearrangement-sensitive 2-arachidonoylglycerol
Roche MJ, et al.
Tetrahedron Letters, 53(30), 3825-3827 (2012)
Tetrahedron Letters, 26, 1411-1411 (1985)
Kang Yuan et al.
Organic letters, 19(6), 1462-1465 (2017-03-08)
We report here a facile B(C6F5)3 catalyzed trans-aminoboration of internal alkynes, furnishing 3-position borylated indoles at ambient temperature. This reaction proceeds with the breaking of a B-N bond and the formation of N-C and B-C bonds to produce indole and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service