Skip to Content
Merck
All Photos(1)

Documents

343528

Sigma-Aldrich

Benzyl sulfone

99%

Synonym(s):

Dibenzyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2SO2
CAS Number:
Molecular Weight:
246.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

SMILES string

O=S(=O)(Cc1ccccc1)Cc2ccccc2

InChI

1S/C14H14O2S/c15-17(16,11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

AWHNUHMUCGRKRA-UHFFFAOYSA-N

Gene Information

Related Categories

General description

Benzyl sulfone is a cyclic sulfone. Benzyl sulfone (Dibenzyl sulfone) on pyrolysis at 290°C yields stillbene and toluene. Mechanism of pyrolysis of dibenzyl sulfones at 600-700°C has been reported. Benzyl sulfone forms sodium salt with metallic sodium and with sodium ethoxide. Cryoscopic behavior of benzyl sulfone was investigated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclic Sulfones. The Condensation of Ethyl Oxalate with Arylmethyl Sulfones.
Overberger CG, et al.
Journal of the American Chemical Society, 72(7), 2856-2859 (1950)
The Cryoscopic Behavior of Organic Compounds in Sulfuric Acid. II. Sulfides, Sulfoxides and Sulfones. Evidence for the Existence of New Sulfur Ions.
Szmant HH and Brost GA.
Journal of the American Chemical Society, 73(9), 4175-4177 (1951)
Pyrolysis of Dibenzyl Sulfones.
Leonard Jr EC.
The Journal of Organic Chemistry, 27(5), 1921-1923 (1962)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service