Skip to Content
Merck
All Photos(1)

Key Documents

331023

Sigma-Aldrich

Nonafluoro-tert-butyl alcohol

99%

Synonym(s):

1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)3COH
CAS Number:
Molecular Weight:
236.04
Beilstein:
1841640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.05

vapor pressure

5.19 psi ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.3 (lit.)

bp

45 °C (lit.)

density

1.693 g/mL at 25 °C (lit.)

SMILES string

OC(C(F)(F)F)(C(F)(F)F)C(F)(F)F

InChI

1S/C4HF9O/c5-2(6,7)1(14,3(8,9)10)4(11,12)13/h14H

InChI key

XZNOAVNRSFURIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonafluoro-tert-butyl alcohol is also referred as perfluorotertiarybutanol and ′artificial blood′.

Application

Nonafluoro-tert-butyl alcohol was employed as a precursor for the preparation of appropriate bis(polyfluoroalkoxymethyl)carbinols and their corresponding mesylates.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclization and rearrangement products from coupling reactions between terminal< i> o</i>-alkynylphenols or< i> o</i>-ethynyl (hydroxymethyl) benzene and 6-halopurines.
Berg TC, et al.
Tetrahedron, 62(25), 6121-6131 (2006)
Greener fluorous chemistry: Convenient preparation of new types of 'CF< sub> 3</sub>-rich'secondary alkyl mesylates and their use for the synthesis of azides, amines, imidazoles and imidazolium salts
Nemes A, et al.
Journal of Fluorine Chemistry, 131(12), 1368-1376 (2010)
Huan Meng et al.
Organic letters, 21(13), 5206-5210 (2019-06-30)
A highly efficient method to incorporate the nonafluoro- tert-butoxy group into various arenes is developed. This C-O cross-coupling reaction proceeds smoothly in the absence of transition-metal catalyst with good functional group tolerance and scalability. In comparison with the conventional approach
Maxime Bourguignon et al.
Macromolecular rapid communications, 42(3), e2000482-e2000482 (2020-10-14)
Polyurethane hydrogels are attractive materials finding multiple applications in various sectors of prime importance; however, they are still prepared by the toxic isocyanate chemistry. Herein the facile and direct preparation in water at room temperature of a large palette of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service