Skip to Content
Merck
All Photos(1)

Key Documents

329894

Sigma-Aldrich

Biphenyl-d10

99 atom % D

Synonym(s):

Diphenyl-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5-C6D5
CAS Number:
Molecular Weight:
164.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

mp

70-72 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])-c2c([2H])c([2H])c([2H])c([2H])c2[2H]

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

ZUOUZKKEUPVFJK-LHNTUAQVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Martín Santos et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1152, 122273-122273 (2020-07-30)
This study reports the development of a method based on headspace (HS)-solid-phase microextraction (SPME)-gas chromatography (GC)-triple quadrupole tandem mass spectrometry (MS/MS) for the quantification of 2- to 4-ring polycyclic aromatic hydrocarbons (PAHs) in saliva samples. Eight unmetabolized compounds (naphthalene, acenaphthylene
Christophe Walgraeve et al.
Analytical and bioanalytical chemistry, 409(1), 335-347 (2016-10-27)
An analytical method was developed and optimized for the quantification of 16 polycyclic aromatic hydrocarbons (PAHs) and 12 oxygenated PAHs in Taxus baccata leaves. Emphasis was given to the development of an in-cell cleanup step using pressurized solvent extraction, a
Banan Soukarieh et al.
Chemosphere, 210, 85-92 (2018-07-10)
This study was carried out in order to investigate the toxicity on Lebanese soil and to show the impact of the anthropogenic activities, industrialization and urbanization, on the release of Polycyclic Aromatic Hydrocarbons (PAHs) in Lebanese soils. Hundred soil samples
Ruixin Xu et al.
The Science of the total environment, 720, 137470-137470 (2020-04-25)
Particulate matter originated from traffic has attracted major interest over the last few years. The semi-volatile organic component of the particles may evaporate with dispersion away from the emission source, creating vapour which may oxidise to form secondary organic aerosol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service