Skip to Content
Merck
All Photos(3)

Key Documents

323438

Sigma-Aldrich

Tetraamminepalladium(II) chloride monohydrate

≥99.99% trace metals basis

Synonym(s):

Tetraamminepalladium(2+) dichloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(NH3)4Cl2·H2O
CAS Number:
Molecular Weight:
263.46
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.99% trace metals basis

form

powder or crystals

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

120 °C (dec.) (lit.)

density

1.91 g/mL at 25 °C (lit.)

SMILES string

N.N.N.N.O.Cl[Pd]Cl

InChI

1S/2ClH.4H3N.H2O.Pd/h2*1H;4*1H3;1H2;/q;;;;;;;+2/p-2

InChI key

WVCXSPJPERKPJS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Tetraamminepalladium (II) chloride monohydrate is used as a precursor to synthesize palladium nanoparticles and Pd (II) complexes.

Application

Tetraamminepalladium(II) chloride monohydrate can be used as a:
  • Precursor for the synthesis of Pd nanoparticles on carbon supports, which is an efficient catalyst for the selective reduction of nitrite to nitrogen.
  • Starting material for the synthesis of a Pd (II) complex which can be immobilized on different substrates to create catalysts with high catalytic activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective reduction of nitrite to nitrogen with carbon-supported Pd--AOT nanoparticles
Perez-Coronado AM, et al.
Journal of Industrial and Engineering Chemistry, 56(41), 11745-11754 (2017)
Immobilization of a Pd (II) complex on hydrophilic graphite oxide and its catalytic investigation in the Heck coupling reaction
Mastalir A, et al.
Applied Organometallic Chemistry, 34(4), e5565-e5565 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service