Skip to Content
Merck
All Photos(1)

Key Documents

32250

Sigma-Aldrich

Diethylene glycol monobutyl ether

≥98.0% (GC)

Synonym(s):

DEGBE, 2-(2-Butoxyethoxy)ethanol, BDG, Butyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
162.23
Beilstein:
1739225
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
technique(s):
GC/GC: suitable
bp:
224-228 °C (lit.)

Assay

≥98.0% (GC)

form

liquid

technique(s)

GC/GC: suitable

impurities

≤0.05% water

refractive index

n20/D 1.432

pH

7

bp

224-228 °C (lit.)

density

0.953 g/mL at 20 °C (lit.)

SMILES string

CCCCOCCOCCO

InChI

1S/C8H18O3/c1-2-3-5-10-7-8-11-6-4-9/h9H,2-8H2,1H3

InChI key

OAYXUHPQHDHDDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylene glycol monobutyl ether (DEGBE) is used:
  • In the preparation of blocked isocyanate systems for coil coating, electro-coating and powder coating.
  • As a cosurfactant in the formation of microemulsions.
  • As a structural component of various liquid crystals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Abegg et al.
Nanomaterials (Basel, Switzerland), 10(6) (2020-06-21)
Nitrogen dioxide (NO2) is a major air pollutant resulting in respiratory problems, from wheezing, coughing, to even asthma. Low-cost sensors based on WO3 nanoparticles are promising due to their distinct selectivity to detect NO2 at the ppb level. Here, we
Doping molecular semiconductors: n-type doping of a liquid crystal perylene diimide.
Gregg BA and Cormier RA
Journal of the American Chemical Society, 123(32), 7959-7960 (2001)
Synthesis and characterization of diethylene glycol monobutyl ether?Blocked diisocyanate crosslinkers.
Ranjbar Z, et al.
Progress in Organic Coatings, 69(4), 426-431 (2010)
Microemulsion systems with monobutyl ether of ethylene glycol or diethylene glycol as cosurfactant.
Chew CH, et al.
Journal of Dispersion Science and Technology, 9(1), 17-31 (1988)
Yeomin Yoon et al.
Journal of membrane science, 261(1-2), 76-86 (2005-09-02)
Rejection characteristics of organic and inorganic compounds were examined for six reverse osmosis (RO) membranes and two nanofiltration (NF) membranes that are commercially available. A batch stirred-cell was employed to determine the membrane flux and the solute rejection for solutions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service